Skip to Content
Merck
All Photos(1)

Key Documents

510866

Sigma-Aldrich

Poly(3-dodecylthiophene-2,5-diyl)

regiorandom

Synonym(s):

P3DDT

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

mol wt

average Mn 30,000
average Mw 70,000

solubility

chloroform, methylene chloride, toluene, and THF: soluble

fluorescence

λex 416 nm; λem 545 nm in chloroform

OPV Device Performance

ITO/PEDOT:PSS/P3DDT/PC61BM (1:3)/LiF/Al

  • Short-circuit current density (Jsc): 2.9 mA/cm2
  • Open-circuit voltage (Voc): 0.6 V
  • Fill Factor (FF): 0.38
  • Power Conversion Efficiency (PCE): 0.65 %

Looking for similar products? Visit Product Comparison Guide

General description

Poly(3-dodecylthiophene-2,5-diyl) (P3DDT) is a poly(3-alkylthiophene) based conducting polymer that shows reversible electrochemistry with high conductivity. It is a regioregular polymer that can be used in the development of a variety of organic electronics based devices.

Application

P3DDT can be used as a conjugating polymer in the fabrication of electrochemical devices, which include polymeric solar cells, chemical sensors and field effect transistors.
This material has solid state properties. Rechargeable battery electrodes, electrochromic devices, chemical and optical sensors, light-emitting diodes, microelectrical amplifiers, field-effect transistors and non-linear optical materials.
Conducting polymer.
Rechargeable battery electrodes, electrochromic devices, chemical and optical sensors, light-emitting diodes, microelectrical amplifiers, field-effect transistors and non-linear optical materials.

Packaging

Packaged in glass bottles

Legal Information

Product of Rieke Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inkjet printed chemical sensor array based on polythiophene conductive polymers
Li B, et al.
Sensors and Actuators B, Chemical, 123(2), 651-660 (2007)
Evaluation of solution-processable carbon-based electrodes for all-carbon solar cells
Ramuz MP, et al.
ACS Nano, 6(11), 10384-10395 (2012)
Journal of the American Chemical Society, 117, 233-233 (1994)
Carbon nanotube network ambipolar field-effect transistors with 108 on/off ratio
Derenskyi V, et al.
Advanced Materials, 26(34), 5969-5975 (2014)
Self-orienting head-to-tail poly (3-alkylthiophenes): new insights on structure-property relationships in conducting polymers
McCullough RD, et al.
Journal of the American Chemical Society, 115(11), 4910-4911 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service