Skip to Content
Merck
All Photos(1)

Documents

541435

Sigma-Aldrich

Poly[2-methoxy-5-(2-ethylhexyloxy)-1,4-phenylenevinylene]

average Mn 70,000-100,000

Synonym(s):

Poly[2-methoxy-5-(2-ethylhexyloxy)-1,4-phenylenevinylene], MEH-PPV

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C18H28O2)n
CAS Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

description

Band gap: 2.3 eV

Quality Level

mol wt

average Mn 70,000-100,000

fluorescence

λex 495 nm; λem 554 nm in toluene

Orbital energy

HOMO -5.3 eV 
LUMO -3 eV 

OLED Device Performance

ITO/PEDOT:PSS/MEH-PPV/Al

  • Color: orange-red
  • Max. Luminance: 220 Cd/m2
  • Max. EQE: 0.3 %
  • Turn-On Voltage: 3.8 V

ITO/PEDOT:PSS/MEH-PPV/PBO/Al
  • Color: orange-red
  • Max. Luminance: 970 Cd/m2
  • Max. EQE: 1.9 %
  • Turn-On Voltage: 2.9 V

ITO/PEDOT:PSS/MEH-PPV/PBZT/Al
  • Color: orange-red
  • Max. Luminance: 1400 Cd/m2
  • Max. EQE: 2.5 %
  • Turn-On Voltage: 2.8 V

OPV Device Performance

ITO/MEH-PPV/BBL/Al

  • Short-circuit current density (Jsc): 1.98 mA/cm2
  • Open-circuit voltage (Voc): 0.93 V
  • Fill Factor (FF): 0.47
  • Power Conversion Efficiency (PCE): 1.1 %

ITO/MEH-PPV:PC61BM(1:4)/Ca
  • Short-circuit current density (Jsc): 2 mA/cm2
  • Open-circuit voltage (Voc): 0.8 V
  • Fill Factor (FF): 0.25
  • Power Conversion Efficiency (PCE): 1.5 %

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Light-emitting conjugated polymer.

Application

Conducting polymer in solar cells and carbon nanotube OLEDs. Useful in producing bright and efficient white polymeric light emitting diodes.
Filter before application.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael Morgen et al.
Pharmaceutical research, 30(1), 257-268 (2012-09-22)
To evaluate using cationic polymeric nanoparticles that interact with hyaluronate to form ionically cross-linked hydrogels to increase the intra-articular retention time of osteoarthritis drugs in the synovial cavity. In vitro tests included nanoparticle release from cross-linked hydrogels using syringe and
Electrochemical In-Situ Conductivity Measurements for Thin Film of Li1-xMn2O4 Spinel
Nishizawa, M.; et al.
Chemistry of Materials, 12, 1367-1371 (2000)
Polybenzobisazoles Are Efficient Electron Transport Materials for Improving the Performance and Stability of Polymer Light-Emitting Diodes
Jenekhe, S. A.; et al.
Chemistry of Materials, 14, 4775 - 4780 (2002)
Polymer Photovoltaic Cells: Enhanced Efficiencies via a Network of Internal Donor-Acceptor Heterojunctions
Yu, G.; et al.
Science, 270, 1789-1791 (1995)
Exciton harvesting, charge transfer, and charge-carrier transport in amorphous-silicon nanopillar/polymer hybrid solar cells
Gowrishankar, V.; Scully, S.; Chan, A.; et al.
Journal of Applied Physics, 103, 064511-064511 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service