Skip to Content
Merck
All Photos(1)

Key Documents

363049

Sigma-Aldrich

Diiodomethane-d2

99 atom % D, contains copper as stabilizer

Synonym(s):

Methylene-d2 iodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD2I2
CAS Number:
Molecular Weight:
269.85
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

99 atom % D

Assay

99% (CP)

contains

copper as stabilizer

bp

68 °C/15 mmHg (lit.)

density

3.325 g/mL at 25 °C (lit.)

mass shift

M+2

SMILES string

[2H]C([2H])(I)I

InChI

1S/CH2I2/c2-1-3/h1H2/i1D2

InChI key

NZZFYRREKKOMAT-DICFDUPASA-N

Related Categories

Application

Used to prepare deuterated fulvene for retro-Diels-Alder studies.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Aldrichimica Acta, 23, 53-53 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service