Skip to Content
Merck
All Photos(1)

Documents

152005

Sigma-Aldrich

Trifluoroacetic acid-d

99.5 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3COOD
CAS Number:
Molecular Weight:
115.03
Beilstein:
1768662
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21

isotopic purity

99.5 atom % D

form

liquid

technique(s)

NMR: suitable

impurities

≤0.500% water
water

refractive index

n20/D 1.3 (lit.)

bp

75 °C (lit.)

mp

-15 °C (lit.)

density

1.493 g/mL at 25 °C (lit.)

mass shift

M+1

SMILES string

[2H]OC(=O)C(F)(F)F

InChI

1S/C2HF3O2/c3-2(4,5)1(6)7/h(H,6,7)/i/hD

InChI key

DTQVDTLACAAQTR-DYCDLGHISA-N

Looking for similar products? Visit Product Comparison Guide

General description

Trifluoroacetic acid-d is a standard purity solvent suitable for routine NMR analyses (conducted at ambient temperatures where quality is less critical).
Trifluoroacetic acid-d is a synthetic reagent having isotopic purity of 99.5atom%D. It undergoes SE′ reaction with (R)-(E)-1,3-diphenyl-3-trimethylsilylpropene, via antistereochemistry to afford (R)-(E)-1,3-diphenylpropene-3-d. Vapor phase dissociation of trifluoroacetic acid-d has been studied.

Application

Trifluoroacetic acid-d may be employed as solvent to investigate the conformations of cyclic peptides by proton NMR spectra. It may be used to compose NMR solvent mixture to investigate the NMR spectra of high molecular weight polyesters such as polyglycolide (PG) and poly(ethylene terephthalate) (PET).

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Conformations of cyclic peptides. IV. Nuclear magnetic resonance studies of cyclo-pentaglycyl-L-leucyl and cyclo-diglycyl-L-histidyldiglycyl-L-tyrosyl.
K D Kopple et al.
Biochemistry, 8(10), 4087-4095 (1969-10-01)
Journal of Labelled Compounds & Radiopharmaceuticals, 28, 25-25 (1990)
Anti stereochemistry in protodesilylation of an optically active allylsilane with trifluoroacetic acid-D.
Hayashi T, et al.
Tetrahedron Letters, 23(44), 4605-4606 (1982)
The Vapor Phase Dissociation of Some Carboxylic Acids. III. 1 Trifluoroacetic Acid and Trifluoroacetic Acid-d2, 3.
Taylor MD and Templeman MB.
Journal of the American Chemical Society, 78(13), 2950-2953 (1956)
Useful NMR solvent mixture for polyesters: Trifluoroacetic acid-d/chloroform-d.
Hariharan R and Pinkus AG.
Polym. Bull., 30(1), 91-95 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service