Skip to Content
Merck
All Photos(1)

Key Documents

67692

Sigma-Aldrich

Iodomethane

purum, ≥99.0% (GC)

Synonym(s):

Methyl iodide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH3I
CAS Number:
Molecular Weight:
141.94
Beilstein:
969135
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

vapor density

4.89 (vs air)

Quality Level

vapor pressure

24.09 psi ( 55 °C)
7.89 psi ( 20 °C)

grade

purum

Assay

≥99.0% (GC)

contains

silver wool as stabilizer

impurities

≤0.5% CH2I2

refractive index

n20/D 1.530
n20/D 1.531 (lit.)

bp

41-43 °C (lit.)
41-43 °C

mp

−64 (lit.)

solubility

water: soluble 14 g/L at 20 °C

density

2.28 g/mL at 25 °C (lit.)

functional group

alkyl halide
iodo

SMILES string

CI

InChI

1S/CH3I/c1-2/h1H3

InChI key

INQOMBQAUSQDDS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Iodomethane (Methyl iodide) is a colorless liquid. It can be prepared by reacting dimethyl sulfate with concentrated KI. Reaction of iodine and phosphorous with methanol has been reported to afford iodomethane.[1] It reacts with tris(triphenylphosphine)chlororhodium(I) to afford the octahedral complex of rhodium(III), [RhlCl(CH3)(PPh3)2(ICH3)].[2] Kinetics of its reaction with atomic chlorine in the gas phase has been investigated. The absolute rate constant of the reaction has been reported.[3]

Application

Iodomethane (Methyl iodide) may be employed in the preparation of stable neutral solutions of hypoiodous acid[4] and various methyl esters.[5]

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 2 - Carc. 2 - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

89.8 °F - closed cup

Flash Point(C)

32.1 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Farabi Temel
Talanta, 211, 120725-120725 (2020-02-20)
This work designs the synthesis of a novel amino morpholine schiff base functionalized calix[4]arene cage (SCC), its deposition onto Quartz Crystal Microbalance (QCM) crystal surface, and usage for the selective detecting of formaldehyde (HCHO). The SCC modified QCM sensor has
Interaction of tris (triphenylphosphine) chlororhodium (I) with iodomethane, methylallyl, and allyl chloride.
Lawson DN, et al.
J. Chem. Soc. Sect. A, 1733-1736 (1966)
Absolute reaction rate of chlorine atoms with iodomethane.
Kambanis KG, et al.
Chemical Physics Letters, 268(5), 498-504 (1997)
Diego Romano Perinelli et al.
Pharmaceutics, 11(6) (2019-06-30)
Quaternary ammonium amphiphiles are a class of compounds with a wide range of commercial and industrial uses. In the pharmaceutical field, the most common quaternary ammonium surfactant is benzalkonium chloride (BAC), which is employed as a preservative in several topical
Eagleson M.
Concise Encyclopedia Chemistry, 649-649 (1994)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service