All Photos(2)
About This Item
Linear Formula:
Dy2O3
CAS Number:
Molecular Weight:
373.00
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Assay
99.9% trace metals basis
form
powder
reaction suitability
reagent type: catalyst
core: dysprosium
density
7.81 g/mL at 25 °C (lit.)
SMILES string
O=[Dy]O[Dy]=O
InChI
1S/2Dy.3O
InChI key
NLQFUUYNQFMIJW-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Dysprosium(III) oxide (Dy₂O₃) is a rare earth metal oxide known for its chemical and thermal stability, high dielectric constant, and wide band gap, making it suitable for various advanced applications. It is widely used in optical coatings for ceramic lamp envelopes, protective layers for optical components, and as a dopant in superconductors. Additionally, Dy₂O₃ has shown promise in enhancing the dielectric and humidity sensing properties of nanomaterials, as well as in radiation shielding applications.
Application
- Structural and optical properties of dysprosium oxide thin films: This article examines the chemical and thermal stability, high dielectric constant, and wide band gap of dysprosium oxide thin films (MF Al-Kuhaili, SMA Durrani, 2014).
- Determination of 8-hydroxy-2′-deoxyguanosine oxidative stress biomarker using dysprosium oxide nanoparticles@ reduced graphene oxide: This research uses dysprosium oxide nanoparticles for the electrochemical oxidation of oxidative stress biomarkers, highlighting its potential in biosensor applications (S Manavalan, U Rajaji, SM Chen, SSP Selvin, 2018).
- Dysprosium doped copper oxide (Cu1-xDyxO) nanoparticles enabled bifunctional electrode for overall water splitting: This study focuses on the synthesis and application of dysprosium-doped copper oxide nanoparticles as bifunctional electrodes for water splitting (JD Rodney, S Deepapriya, MC Robinson, 2021).
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
J M Peeters et al.
Physics in medicine and biology, 51(6), N127-N137 (2006-03-03)
Susceptibility markers for passive tracking need to be small in order to maintain the shape and mechanical properties of the endovascular device. Nevertheless, they also must have a high magnetic moment to induce an adequate artefact at a variety of
Masoud Salavati-Niasari et al.
Ultrasonics sonochemistry, 17(5), 870-877 (2010-03-23)
Dysprosium carbonates nanoparticles were synthesized by the reaction of dysprosium acetate and NaHCO(3) by a sonochemical method. Dysprosium oxide nanoparticles with average size about 17 nm were prepared from calcination of Dy(2)(CO(3))(3).1.7H(2)O nanoparticles. Dy(OH)(3) nanotubes were synthesized by sonication of
S E Seltzer et al.
Journal of computer assisted tomography, 5(3), 370-374 (1981-06-01)
We used a stepwise approach to identify, design, synthesize, and test new high atomic number particulate contrast agents that would be especially well suited for use with computed tomography (CT). Our goal was to produce extremely radiopaque compounds with highly
B Vengala Rao et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 71(3), 951-953 (2008-04-01)
Emission spectrum of Dy(3+):Ca(4)GdO(BO(3))(3) powder phosphor has been analyzed. Two emission bands at 492 and 588 nm with lambda(exci)=311 nm ((6)H(15/2)-->(4)L(19/2)) have been measured from this phosphor. For the intense yellow emission at 588 nm ((4)F(9/2)-->(6)H(13/2)), its lifetime has been
Happy et al.
Journal of nanoscience and nanotechnology, 7(3), 907-915 (2007-04-25)
The experimental parameters that control the size and size distribution of dysprosium oxide nanoparticles synthesized by homogeneous precipitation technique have been systematically investigated. The particles were characterized with respect to their size, shape, and thermal decomposition behavior. It was found
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service