Skip to Content
Merck
All Photos(2)

Key Documents

203238

Sigma-Aldrich

Erbium(III) oxide

≥99.99% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Er2O3
CAS Number:
Molecular Weight:
382.52
EC Number:
MDL number:
UNSPSC Code:
12352303
eCl@ss:
38161402
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

≥99.99% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: erbium

density

8.64 g/mL at 25 °C (lit.)

SMILES string

O=[Er]O[Er]=O

InChI

1S/2Er.3O

InChI key

VQCBHWLJZDBHOS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Erbium(III) oxide (Er₂O₃) is a metal oxide with high thermal stability, optical transparency, and excellent luminescent characteristics. It is widely used in microelectronics, optoelectronics, and biomedical fields. The material is particularly valued for its role in solid-state lasers and optical amplifiers. Additionally, Er₂O₃ is utilized in the production of high-refractive index glasses for optical devices and composite electrolyte for fuel cells.

Application

  • Green emission of tellurite based glass containing erbium oxide nanoparticles: This research explores the optical properties of erbium oxide nanoparticles in glass, which is important for academia chemists studying photonic materials (Noorazlan et al., 2016).
  • … shielding properties for Oxide Dispersion-Strengthened (ODS) alloys through Erbium (III) oxide, Samarium (III) oxide, and Praseodymium (III) oxide into 316L matrix: Relevant for material science, this research investigates the enhancement of shielding properties in alloys by incorporating erbium oxide (Güler et al., 2024).
  • Electrochemical synthesis and characterization of erbium oxide: This paper discusses the synthesis and potential applications of erbium oxide in various high-tech industries, relevant for both academia and industry researchers focusing on advanced material applications (Lee and Kim, 2016).

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Priyam Singh et al.
Nanotechnology, 28(41), 415701-415701 (2017-07-19)
We present novel inorganic-organic hybrid nanoparticles (HNPs) constituting inorganic NPs, NaY
Liu Peng et al.
Biological trace element research, 104(3), 275-284 (2005-06-03)
A microcalorimetric technique was used to evaluate the influence of Er3+ on Halobacterium halobium R1 growth. By means of a LKB-2277 Bioactivity Monitor ampoule method, we obtained the thermogenic curves of H. halobium R1 growth at 37 degrees C. In
J J Borm et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 36(5), 888-892 (1995-05-01)
The feasibility of postproduction neutron activation of an enteric-coated pancreatic enzyme preparation for in vivo gastric emptying studies has been investigated. During production of this multicomponent preparation, small amounts of 170Er-enriched erbium oxide, suitable for neutron activation, were added. Postproduction
Darya Radziuk et al.
Langmuir : the ACS journal of surfaces and colloids, 27(23), 14472-14480 (2011-10-26)
Ultrasound (20 kHz, 29 W·cm(-2)) is employed to form three types of erbium oxide nanoparticles in the presence of multiwalled carbon nanotubes as a template material in water. The nanoparticles are (i) erbium carboxioxide nanoparticles deposited on the external walls
Shixun Dai et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 62(1-3), 431-437 (2005-11-01)
(100-x)TeO(2)-xNb(2)O(5) (x=5-20) niobic tellurite glasses doped with 0.5 mol.% Er(2)O(3) were synthesized, and their thermal, mechanical, and spectroscopic properties were measured and compared to the properties of the typical 75TeO(2)-20ZnO-5Na(2)O (TZN) tellurite glass. The refractive index (n(d)), density (rho), and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service