Skip to Content
Merck
All Photos(2)

Key Documents

289167

Sigma-Aldrich

Thulium(III) oxide

99.9% trace metals basis

Synonym(s):

Thulia, Thulium oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Tm2O3
CAS Number:
Molecular Weight:
385.87
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

99.9% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: thulium

SMILES string

O=[Tm]O[Tm]=O

InChI

1S/3O.2Tm

InChI key

CWEYWFRFCUNVEY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B E Lambert et al.
Radiation and environmental biophysics, 20(1), 11-20 (1981-01-01)
The short term effects on the free macrophage population in the mouse lung, following inhalation of thulium-170 oxide, have been studied, with a view to using the system as a model with which to compare the short term effects of
G X Chen et al.
Journal of fluorescence, 17(3), 301-307 (2007-03-30)
We report on 2.0-microm emission characteristic and energy transfer of Ho3+-doped tellurite glass sensitized by Tm3+ upon excitation of a conventional 808 nm laser diode. The Judd-Ofelt strength parameters, spontaneous radiative transition probabilities and radiative lifetime of Ho3+ have been
B Chen et al.
Guang pu xue yu guang pu fen xi = Guang pu, 21(3), 287-289 (2003-09-02)
In this work, we have a designation and preparation of MFT glasses for upconversion, the glasses consisted of TeO2 and fluoride: PbF2, AlF3, BaF2, NaF and the impurity Tm2O3. In this glass system the oxide improve forming ability, the fluorides
Zhengze Yu et al.
ACS nano, 9(11), 11064-11074 (2015-10-13)
Photodynamic therapy (PDT) is a well-established modality for cancer therapy, which locally kills cancer cells when light irradiates a photosensitizer. However, conventional PDT is often limited by the extremely short lifespan and severely limited diffusion distance of reactive oxygen species

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service