Skip to Content
Merck
All Photos(1)

Documents

289221

Sigma-Aldrich

Europium(III) oxide

99.9% trace metals basis

Synonym(s):

Europia, Europium oxide, Europium trioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Eu2O3
CAS Number:
Molecular Weight:
351.93
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: europium

density

7.42 g/mL at 25 °C (lit.)

SMILES string

O=[Eu]O[Eu]=O

InChI

1S/2Eu.3O

InChI key

RSEIMSPAXMNYFJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Europium(III) oxide, also known as europia, is a rare earth metal oxide with high density of 7.42 g/mL at 25 °C. It is widely used in the production of phosphors for cathode ray tubes, X-ray intensifying screens, and fluorescent lamps due to its strong red emission under UV excitation. Europium(III) oxide is a convenient source of europium for doping other metal oxides. Europium doping often imparts fluorescence and photoluminescence, owing to europium’s electron configuration with partially filled f-orbitals. Our high-purity Eu2O3 is a key component in the development of energy-efficient lighting and displays, including LED-based technologies.

Application

  • Microwave-assisted synthesis of europium (III) oxide decorated reduced graphene oxide nanocomposite for detection of chloramphenicol in food samples: Describes the use of Eu2O3 nanoparticles decorated reduced graphene oxide for developing an amperometric sensor for chloramphenicol detection (Rajaji et al., 2019).

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Islam Mohamed Abdelmonem et al.
International journal of biological macromolecules, 164, 2258-2266 (2020-08-18)
Removal behaviors of 152+154Eu, 60Co, and 134Cs radionuclides onto Chitosan-acrylic acid-1-vinyl-2-vinylpyrrolidone/oxidized multi-walled carbon nanotubes (CTS-AA-VP/o-MWCNTs) composite has been investigated by batch adsorption technique. CTS-AA-VP/o-MWCNTs composite has been synthesized by copolymerization of acrylic acid (AA) and 1-vinyl-2-vinylpyrrolidone (VP) onto the surface

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service