Skip to Content
Merck
All Photos(1)

Documents

282650

Sigma-Aldrich

D-Glucose-6,6-d2

≥98 atom % D, ≥99% (CP)

Synonym(s):

Deuterated Glucose, Labeled Glucose, Dextrose-6,6-d2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6D2H10O6
CAS Number:
Molecular Weight:
182.17
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

≥98 atom % D

Quality Level

Assay

≥99% (CP)

form

powder

mp

150-152 °C (lit.)

mass shift

M+2

SMILES string

[2H]C([2H])(O)[C@H]1OC(O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5-,6?/m1/s1/i1D2

InChI key

WQZGKKKJIJFFOK-VARZTKRMSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nadejda Bozadjieva Kramer et al.
Cell reports, 33(2), 108239-108239 (2020-10-15)
Elevated levels of branched-chain amino acids (BCAAs) and their metabolites are strongly positively associated with obesity, insulin resistance, and type 2 diabetes. Bariatric surgery is among the best treatments for weight loss and associated morbidities. Clinical studies have reported that
Boyoung Han et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1160, 122370-122370 (2020-09-20)
Monosaccharide composition of biological samples can reflect an individual's health status. Monitoring the concentration of individual monosaccharides in human serum requires a technique for the simultaneous analysis of multiple monosaccharide molecules. Furthermore, certified reference materials (CRMs) for overall monosaccharide composition
Erwei Li et al.
Cell metabolism, 30(2), 319-328 (2019-06-25)
Asprosin is a fasting-induced hormone that promotes glucose production in the liver and stimulates appetite in the hypothalamus by activating the cAMP signaling pathway via an unknown G protein-coupled receptor (GPCR). However, the bona fide receptor of Asprosin is unclear.
Wen-Qiang Wu et al.
Nucleic acids research, 43(9), 4614-4626 (2015-04-22)
Mutations in the RecQ DNA helicase gene BLM give rise to Bloom's syndrome, which is a rare autosomal recessive disorder characterized by genetic instability and cancer predisposition. BLM helicase is highly active in binding and unwinding G-quadruplexes (G4s), which are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service