Skip to Content
Merck
All Photos(3)

Key Documents

372382

Sigma-Aldrich

Sodium bicarbonate-13C

98 atom % 13C, 99% (CP)

Synonym(s):

13C Labeled sodium bicarbonate, Sodium hydrogencarbonate-13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaH13CO3
CAS Number:
Molecular Weight:
85.00
MDL number:
UNSPSC Code:
51113400
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % 13C

Quality Level

Assay

99% (CP)

form

white powder

mp

>300 °C (lit.)

mass shift

M+1

storage temp.

room temp

SMILES string

[Na].O[13C](O)=O

InChI

1S/CH2O3.Na/c2-1(3)4;/h(H2,2,3,4);/q;+1/p-1/i1+1;

InChI key

UIIMBOGNXHQVGW-YTBWXGASSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium hydrogencarbonate-13C is isotopically enriched compound of sodium bicarbonate.

Application

Sodium bicarbonate-13C can be used:
  • As a probe for NMR− and MRI− based pH imaging.
  • To predict the reaction mechanism in selective hydrogenation of bicarbonate to formate, catalyzed by a ruthenium(III) complex.
  • For 13C labeling of amino acid sample for compound-specific stable isotope analysis (CSIA).

Sodium bicarbonate-13C is used to study the lactate metabolism by using hyperpolarized 13C pH imaging and multiparametric MRI.This reagent can be used as probe in nuclear magnetic resonance spectroscopy studies, which enabled large signal gains for low-toxicity hyperpolarized 13C pH imaging in a phantom and in vivo in a murine model of prostate cancer.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

RuIII (edta) catalyzed hydrogenation of bicarbonate to formate.
Chatterjee D and Sarkar P
Journal of Coordination Chemistry, 69(4), 650-655 (2016)
The influence of hydrolysis and derivatization on the determination of amino acid content and isotopic ratios in dual-labeled (13C, 15N) white clover.
Enggrob KL, et al.
Rapid Communications in Mass Spectrometry, 33(1), 21-30 (2019)
David E Korenchan et al.
Oncotarget, 10(58), 6096-6110 (2019-11-07)
There is an unmet clinical need for new and robust imaging biomarkers to distinguish indolent from aggressive prostate cancer. Hallmarks of aggressive tumors such as a decrease in extracellular pH (pHe) can potentially be used to identify aggressive phenotypes. In
Ferdia A Gallagher et al.
NMR in biomedicine, 24(8), 1006-1015 (2011-08-04)
pH is a fundamental physiological parameter that is tightly controlled by endogenous buffers. The acid-base balance is altered in many disease states, such as inflammation, ischemia and cancer. Despite the importance of pH, there are currently no routine methods for
Quantified pH imaging with hyperpolarized 13 C-bicarbonate
Scholz DJ, et al.
Magnetic Resonance in Medicine : Official Journal of the Society of Magnetic Resonance in Medicine / Society of Magnetic Resonance in Medicine, 73(6), 2274-2282 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service