Skip to Content
Merck
All Photos(6)

Key Documents

389374

Sigma-Aldrich

D-Glucose-13C6

≥99 atom % 13C, ≥99% (CP)

Synonym(s):

Labeled Glucose, D-Glucose-ul-13C, Dextrose-13C6

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C6H12O6
CAS Number:
Molecular Weight:
186.11
MDL number:
UNSPSC Code:
12142200
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

≥99 atom % 13C

Assay

≥99% (CP)

form

powder

optical activity

[α]25/D +52.0°, c = 2 in H2O (trace NH4OH)

technique(s)

bio NMR: suitable
protein expression: suitable
protein purification: suitable

mp

150-152 °C (lit.)

mass shift

M+6

SMILES string

O[13CH2][13C@H]1O[13C@H](O)[13C@H](O)[13C@@H](O)[13C@@H]1O

InChI

1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5-,6?/m1/s1/i1+1,2+1,3+1,4+1,5+1,6+1

InChI key

WQZGKKKJIJFFOK-NYKRWLDMSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

D-Glucose-13C6 has been used:
  • for de novo fatty acid (FA) synthesis
  • as a component in tracing media for the analysis of stable isotope tracer
  • in minimal media for recombinant protein production
  • for cell suspension

Biochem/physiol Actions

D-Glucose-13C6 functions as a tracer and helps to access glucose kinetics in vivo.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Oda et al.
Biochemistry, 33(17), 5275-5284 (1994-05-03)
All of the individual carboxyl groups (the side-chain carboxyl groups of Asp and Glu, and the C-terminal alpha-carboxyl group) in Escherichia coli ribonuclease HI, which is an enzyme that cleaves the RNA strand of a RNA/DNA hybrid, were pH-titrated, and
Metabolic profiling reveals a coordinated response of isolated lamb's (Valerianella locusta, L.) lettuce cells to sugar starvation and low oxygen stress
Victor BMM, et al.
Postharvest biology and technology, 126, 23-33 (2017)
Zoran Dinev et al.
Carbohydrate research, 341(10), 1743-1747 (2006-04-11)
A simple gram-scale synthesis of uridine diphospho(13C6)glucose is presented from D-(13C6)glucose. The critical step uses a 1H-tetrazole-catalyzed coupling of 2,3,4,6-tetra-O-acetyl-alpha-D-glucopyranosyl-1-phosphate and UMP-morpholidate. The uridine diphospho(13C6)glucose was used in the structural identification of (1-->3)-beta-D-glucan from Lolium multiflorum.
RSK Regulates PFK-2 Activity to Promote Metabolic Rewiring in Melanoma
Houles T, et al.
Cancer Research, canres-ca2215 (2018)
Proceedings: Teratogenic actions of C13-glucose on the early development of the mouse embryo.
H Spielman et al.
Naunyn-Schmiedeberg's archives of pharmacology, 287 Suppl, R86-R86 (1975-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service