Merck
OLFR734 Mediates Glucose Metabolism as a Receptor of Asprosin.
Cell metabolism (2019-06-25)
Erwei Li, Haili Shan, Liqun Chen, Aijun Long, Yuanyuan Zhang, Yang Liu, Liangjie Jia, Fangchao Wei, Jinbo Han, Tong Li, Xiaohui Liu, Haiteng Deng, Yiguo Wang
ABSTRACT
Asprosin is a fasting-induced hormone that promotes glucose production in the liver and stimulates appetite in the hypothalamus by activating the cAMP signaling pathway via an unknown G protein-coupled receptor (GPCR). However, the bona fide receptor of Asprosin is unclear. Here, we have identified that the olfactory receptor OLFR734 acts as a receptor of Asprosin to modulate hepatic glucose production. Olfr734 knockout mice show a blunted response to Asprosin, including attenuated cAMP levels and hepatic glucose production, and improved insulin sensitivity. As Olfr734 deficiency dramatically attenuates both fasting and high-fat-diet-induced glucose production, our results demonstrate a critical role of OLFR734 as a receptor of Asprosin to maintain glucose homeostasis during fasting and in obesity.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
Sigma-Aldrich
Forskolin, For use in molecular biology applications
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture