Skip to Content
Merck
All Photos(3)

Key Documents

212903

Sigma-Aldrich

Terbium(III) chloride hexahydrate

99.9% trace metals basis

Synonym(s):

Terbium chloride hexahydrate, Terbium trichloride hexahydrate, Trichloroterbium hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TbCl3 · 6H2O
CAS Number:
Molecular Weight:
373.38
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
solid

grade

for analytical purposes

Assay

99.9% trace metals basis

form

solid

reaction suitability

reagent type: catalyst
core: terbium

impurities

≤1500.0 ppm Trace Rare Earth Analysis

density

4.35 g/mL at 25 °C (lit.)

SMILES string

O.O.O.O.O.O.Cl[Tb](Cl)Cl

InChI

1S/3ClH.6H2O.Tb/h3*1H;6*1H2;/q;;;;;;;;;+3/p-3

InChI key

ULJUVCOAZNLCJZ-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Terbium(III) chloride hexahydrate can be used to study the effect of Tb(III) ions on the micellization properties of surfactants.

It can also be used as aprecursor to synthesize a label-freeaptasensor for the detection of ofloxacin (OFL) residues in the food.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sébastien Moret et al.
Forensic science international, 291, 83-93 (2018-09-02)
Metal-Organic Frameworks (MOFs) are porous crystalline structures, currently used as sensors, separation membranes, and as catalysts. Due to their physicochemical and optical properties, they have been recently proposed for fingermark detection. This study further explored their potential for fingermark detection.
Eitam Vinegrad et al.
ACS nano, 13(1), 601-608 (2018-12-07)
The occurrence of biological homochirality is attributed to symmetry-breaking mechanisms which are still debatable. Studies of symmetry breaking require tools for monitoring the population ratios of individual chiral nano-objects, such as molecules, polymers, or nanocrystals. Moreover, mapping their spatial distributions
Uri Hananel et al.
Proceedings of the National Academy of Sciences of the United States of America, 116(23), 11159-11164 (2019-05-18)
Symmetry plays a crucial part in our understanding of the natural world. Mirror symmetry breaking is of special interest as it is related to life as we know it. Studying systems which display chiral amplification, therefore, could further our understanding
A Shinohara et al.
Toxicology, 66(1), 93-103 (1991-02-11)
To investigate the biological effects of terbium (Tb), male mice were intraperitoneally administered with TbCl3 at doses of 10, 50, or 250 mg Tb/kg. The Tb distribution in organs was determined after 18-20 h of injection by using a spectrofluorometer.
H Shimada et al.
Journal of toxicology and environmental health, 48(1), 81-92 (1996-05-01)
Terbium (Tb) is a rare earth metal that finds use in several emerging technologies. However, little is known about the biological effects of Tb. Thus, in this study the pulmonary toxicity of systemic Tb in mice was investigated. Mice were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service