Skip to Content
Merck
All Photos(2)

Key Documents

217212

Sigma-Aldrich

Terbium(III) nitrate hexahydrate

99.999% trace metals basis

Synonym(s):

Terbium(3+) trinitrate hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Tb(NO3)3 · 6H2O
CAS Number:
Molecular Weight:
453.03
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
crystals and lumps

grade

for analytical purposes

Assay

99.999% trace metals basis

form

crystals and lumps

reaction suitability

reagent type: catalyst
core: terbium

impurities

≤20.0 ppm Trace Rare Earth Analysis

SMILES string

O.O.O.O.O.O.[Tb+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/3NO3.6H2O.Tb/c3*2-1(3)4;;;;;;;/h;;;6*1H2;/q3*-1;;;;;;;+3

InChI key

UTCARTSNNKGRTD-UHFFFAOYSA-N

Application

Forms a permanently microporous material [Tb2(1,4-benzenedicarboxylate)3] whose luminescence properties hold promise for sensor applications.

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Balwinder Kaur et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 214, 261-268 (2019-02-21)
Highly crystalline polyethylene glycol (PEG) coated Tb3+ doped ZnS nanoparticles have been synthesized and successfully used for norfloxacin sensing. The crystallographic and morphological analyses of PEG coated Tb3+ doped ZnS nanoparticles were performed by X-ray diffraction and Transmission electron microscopy
Svetlana V Fedorenko et al.
Materials science & engineering. C, Materials for biological applications, 76, 551-558 (2017-05-10)
The work introduces Tb(III)-centered luminescence of amino-modified silica nanoparticles doped with Tb(III) complexes for cellular imaging. For these purposes water-in-oil procedure was optimized for synthesis of 20 and 35nm luminescent nanoparticles with amino-groups embedded on the surface. The obtained results
Reineke, T.M. et al.
Journal of the American Chemical Society, 121, 1651-1651 (1999)
Svetlana V Fedorenko et al.
Journal of inorganic biochemistry, 182, 170-176 (2018-02-28)
The present work introduces composite luminescent nanoparticles (Ag0-Tb3+-SNs), where ultra-small nanosilver (4 ± 2 nm) is deposited onto amino-modified silica nanoparticles (35±6 nm) doped by green luminescent Tb(III) complexes. Ag0-Tb3+-SNs are able to image cancer (Hep-2) cells in confocal microscopy measurements due to
Elisabetta Fanizza et al.
International journal of molecular sciences, 20(13) (2019-06-30)
The low photostability of conventional organic dyes and the toxicity of cadmium-based luminescent quantum dots have prompted the development of novel probes for in vitro and in vivo labelling. Here, a new fluorescent lanthanide probe based on silica nanoparticles is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service