Skip to Content
Merck
All Photos(1)

Documents

429732

Sigma-Aldrich

Europium(III) chloride

anhydrous, powder, 99.99% trace metals basis

Synonym(s):

Europium trichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
EuCl3
CAS Number:
Molecular Weight:
258.32
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

grade

anhydrous

Assay

99.99% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: europium

impurities

≤150.0 ppm Trace Rare Earth Analysis

mp

850 °C (lit.)

density

4.89 g/mL at 25 °C (lit.)

SMILES string

Cl[Eu](Cl)Cl

InChI

1S/3ClH.Eu/h3*1H;/q;;;+3/p-3

InChI key

NNMXSTWQJRPBJZ-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

  • Photochemical Recovery of Europium from Non-aqueous Solutions: This study explores the photochemical processes involving Europium(III) chloride, which are significant for applications in material recovery and recycling (B Van den Bogaert, L Gheeraert, 2016).
  • Electrochemistry of Europium(III) Chloride in Various Eutectics: Investigation of the electrochemical properties of Europium(III) chloride in different molten salt mixtures, important for developing new materials and nuclear applications (CA Schroll, S Chatterjee, TG Levitskaia, 2017).

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Longjun Wang et al.
Steroids, 89, 41-46 (2014-08-06)
An ultrasensitive and selective method is presented for the determination of diethylstilbestrol (DES) using time-resolved fluorescence immunoassay (TRFIA) based on double-codified gold nanoparticles (DC-AuNPs). In this system, the DC-AuNPs, that are gold nanoparticles (AuNPs) modified with anti-DES antibody and SH-dsDNA-biotin
Yoshiko Fujita et al.
Environmental science & technology, 49(16), 9460-9468 (2015-07-02)
Increasing rare earth element (REE) supplies by recycling and expanded ore processing will result in generation of new wastewaters. In some cases, disposal to a sewage treatment plant may be favored, but plant performance must be maintained. To assess the
André M Bienfait et al.
Dalton transactions (Cambridge, England : 2003), 43(46), 17324-17332 (2014-10-17)
Trivalent Eu[N(SiHMe2)2]3(THF)2 can easily be synthesized by applying a routine salt metathesis protocol (EuCl3(THF)2 and 3 equiv. of Li[N(SiHMe2)2] in n-hexane) which crystallizes isotypically to its analogues of the rare-earth metal series (space group P21/c). Transsilylamination of Eu[N(SiMe3)2]2(THF)2 with a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service