Skip to Content
Merck
All Photos(3)

Key Documents

204560

Sigma-Aldrich

Terbium(III) chloride hexahydrate

99.999% trace metals basis

Synonym(s):

Terbium trichloride hexahydrate, Terbium(3+) trichloride hexahydrate, Trichloroterbium hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TbCl3 · 6H2O
CAS Number:
Molecular Weight:
373.38
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder and chunks

grade

for analytical purposes

Assay

99.999% trace metals basis

form

powder and chunks

reaction suitability

reagent type: catalyst
core: terbium

impurities

≤15.0 ppm Trace Rare Earth Analysis

density

4.35 g/mL at 25 °C (lit.)

SMILES string

O.O.O.O.O.O.Cl[Tb](Cl)Cl

InChI

1S/3ClH.6H2O.Tb/h3*1H;6*1H2;/q;;;;;;;;;+3/p-3

InChI key

ULJUVCOAZNLCJZ-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Shinohara et al.
Toxicology, 66(1), 93-103 (1991-02-11)
To investigate the biological effects of terbium (Tb), male mice were intraperitoneally administered with TbCl3 at doses of 10, 50, or 250 mg Tb/kg. The Tb distribution in organs was determined after 18-20 h of injection by using a spectrofluorometer.
H Shimada et al.
Journal of toxicology and environmental health, 48(1), 81-92 (1996-05-01)
Terbium (Tb) is a rare earth metal that finds use in several emerging technologies. However, little is known about the biological effects of Tb. Thus, in this study the pulmonary toxicity of systemic Tb in mice was investigated. Mice were
David L Rosen
Applied optics, 45(13), 3152-3157 (2006-04-28)
An experimental investigation is described of a terbium luminescence sensor for continuous collection and detection of bacterial endospores suspended in the atmosphere. A sensor is presented that continuously monitors and rapidly detects small concentrations of aerosolized bacterial endospores. The aerosol
J L Cox et al.
Melanoma research, 12(3), 211-219 (2002-07-26)
Alterations in the extracellular Ca(2+) or K(+) concentration had significant influences on the motility of B16F10 melanoma cells measured in the absence of exogenous integrins using a conventional Boyden chamber assay. At normal K(+) concentrations, motility increased slightly when the
William J Rieter et al.
Journal of the American Chemical Society, 130(35), 11584-11585 (2008-08-09)
Pt-containing nanoscale coordination polymer (NCP) particles with the formula of Tb2(DSCP)3(H2O)12 (where DSCP represents disuccinatocisplatin), NCP-1, were precipitated from an aqueous solution of Tb3+ ions and DSCP bridging ligands via the addition of a poor solvent. SEM and TEM images

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service