All Photos(1)
About This Item
Linear Formula:
CH3P(S)(OC2H5)2
CAS Number:
Molecular Weight:
168.19
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
form
liquid
refractive index
n20/D 1.464 (lit.)
bp
76-79 °C/13 mmHg (lit.)
density
1.055 g/mL at 25 °C (lit.)
SMILES string
CCOP(C)(=S)OCC
InChI
1S/C5H13O2PS/c1-4-6-8(3,9)7-5-2/h4-5H2,1-3H3
InChI key
GEDUXIUTPVHAAV-UHFFFAOYSA-N
Related Categories
General description
O,O′-Diethyl methylphosphonothioate is a phosphorous containing organic building block. Structure of the O,O′-diethyl methylphosphonothioate has been investigated by high resolution NMR.
Signal Word
Warning
Hazard Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
165.2 °F - closed cup
Flash Point(C)
74 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
STRUCTURE OF O, O'-DIETHYL METHYLPHOSPHONOTHIOATE AND CONJUGATIVE PROPERTIES OF THE P [UNK] S BOND.
Finegold H.
Journal of the American Chemical Society, 82(10), 2641-2643 (1960)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service