Skip to Content
Merck
All Photos(3)

Documents

115606

Sigma-Aldrich

Hexanal

98%

Synonym(s):

Aldehyde C6, Caproaldehyde, Hexyl aldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CHO
CAS Number:
Molecular Weight:
100.16
Beilstein:
506198
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39021112
PubChem Substance ID:
NACRES:
NA.22

vapor density

>1 (vs air)

Quality Level

vapor pressure

10 mmHg ( 20 °C)

Assay

98%

refractive index

n20/D 1.4035 (lit.)

bp

130-131 °C (lit.)

density

0.815 g/mL at 25 °C (lit.)
0.834 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CCCCCC=O

InChI

1S/C6H12O/c1-2-3-4-5-6-7/h6H,2-5H2,1H3

InChI key

JARKCYVAAOWBJS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hexanal is present in lactoperoxidase-bleached WPC80 (whey protein concentrate with 80% protein).

Application

Hexanal has been used in the quantification of carbonyls that contribute to the complex flavor of olive oil. It can also be used for Wittig and aldol reactions.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

77.0 °F - closed cup

Flash Point(C)

25 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R E Campbell et al.
Journal of dairy science, 96(12), 7404-7413 (2013-10-22)
Chemical bleaching of fluid whey and retentate with hydrogen peroxide (HP) alone requires high concentrations (100-500 mg of HP/kg) and recent studies have demonstrated that off-flavors are generated during chemical bleaching that carry through to spray-dried whey proteins. Bleaching of
Hanjiang Zhu et al.
Journal of agricultural and food chemistry, 61(50), 12253-12259 (2013-11-28)
The enzymatic and chemical oxidation reaction in olive oil produces many volatile carbonyl compounds that contribute to the complex flavor of olive oil. A novel ultrahigh performance liquid chromatography (UHPLC) method with dynamic headspace sampling and 2,4-dinitrophenylhydrazine (DNPH) derivatization were
Juhun Park et al.
The Analyst, 137(14), 3249-3254 (2012-04-13)
We developed an olfactory-nanovesicle-fused carbon-nanotube-transistor biosensor (OCB) that mimics the responses of a canine nose for the sensitive and selective detection of hexanal, an indicator of the oxidation of food. OCBs allowed us to detect hexanal down to 1 fM
Laura Purriños et al.
Meat science, 93(2), 344-350 (2012-10-30)
The volatile profile of dry-cured "lacón" that has been inoculated with three different yeasts were determined and compared with a non-inoculated dry-cured "lacón". Yeasts (Debaryomyces hansenii, Candida deformans and Candida zeylanoides) that were used as starter cultures in the present
Nicoletta Faraone et al.
Insects, 11(8) (2020-08-08)
Blacklegged ticks, Ixodes scapularis, represent a significant public health concern due to their vectoring of tick-borne disease. Despite their medical importance, there is still limited knowledge of the chemosensory system of this species, and thus a poor understanding of host-seeking

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service