Skip to Content
Merck
All Photos(1)

Documents

73117

Supelco

1-Hexanol

analytical standard

Synonym(s):

Hexyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5OH
CAS Number:
Molecular Weight:
102.17
Beilstein:
969167
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

4.5 (vs air)

vapor pressure

1 mmHg ( 25.6 °C)

Assay

≥99.9% (GC)

autoignition temp.

559 °F

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.418 (lit.)
n20/D 1.418

bp

156-157 °C (lit.)

mp

−52 °C (lit.)

density

0.814 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCCCCCO

InChI

1S/C6H14O/c1-2-3-4-5-6-7/h7H,2-6H2,1H3

InChI key

ZSIAUFGUXNUGDI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Hexanol is an organic alcohol, which has application in the synthesis of antiseptics, fragrances, perfumes, etc. It is also used as a component of plasticizers.

Application

1-Hexanol may be used as an analytical standard for the determination of the analyte in wine, plant samples, flowers, fruits and oil samples by various chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

140.0 °F - closed cup

Flash Point(C)

60 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Profiling the chemical content of Opuntia ficus-indica flowers by HPLC-PDA-ESI-MS and GC/EIMS analyses
De Leo M, et al.
Phytochemistry Letters, 3(1), 48-52 (2010)
On-line gas chromatography combustion/pyrolysis isotope ratio mass spectrometry (HRGC-C/P-IRMS) of major volatiles from pear fruit (Pyrus communis) and pear products
Kahle K, et al.
Food Chemistry, 91(3), 449-455 (2005)
J Ruther
Journal of chromatography. A, 890(2), 313-319 (2000-09-29)
A series of ubiquitously occurring saturated and monounsaturated six-carbon aldehydes, alcohols and esters thereof is summarised as 'green leaf volatiles' (GLVs). The present study gives a comprehensive data collection of retention indices of 35 GLVs on commonly used non-polar DB-5
Patty?s toxicology, 6 volume set
Patty's Toxicology (2012)
C Ortega et al.
Journal of chromatography. A, 923(1-2), 205-214 (2001-08-21)
A method for the simultaneous determination of major (10-200 mg/l) and minor (0.1-10 mg/l) volatile compounds from wine has been optimised and validated. A 3-ml volume of wine is diluted with water (7 ml), salted with 4.5 g of ammonium

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service