Skip to Content
Merck
All Photos(3)

Key Documents

W255726

Sigma-Aldrich

Hexanal

greener alternative

natural, ≥95%, FG

Synonym(s):

Aldehyde C6, Caproaldehyde, Hexyl aldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CHO
CAS Number:
Molecular Weight:
100.16
FEMA Number:
2557
Beilstein:
506198
EC Number:
Council of Europe no.:
96c
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
5.008
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
natural
food allergen:
no known allergens

grade

FG
Fragrance grade
Halal
Kosher
natural

Quality Level

300
400

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

vapor density

>1 (vs air)

vapor pressure

10 mmHg ( 20 °C)

Assay

≥95%

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.4035 (lit.)

bp

130-131 °C (lit.)

density

0.815 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

greener alternative category

Organoleptic

fatty; green; fruity

storage temp.

2-8°C

SMILES string

CCCCCC=O

InChI

1S/C6H12O/c1-2-3-4-5-6-7/h6H,2-5H2,1H3

InChI key

JARKCYVAAOWBJS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product is a Biobased products, showing key improvements in Green Chemistry Principles “Less Hazardous Chemical Syntheses” and “Use of Renewable Feedstock”.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

77.0 °F - closed cup

Flash Point(C)

25 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juhun Park et al.
The Analyst, 137(14), 3249-3254 (2012-04-13)
We developed an olfactory-nanovesicle-fused carbon-nanotube-transistor biosensor (OCB) that mimics the responses of a canine nose for the sensitive and selective detection of hexanal, an indicator of the oxidation of food. OCBs allowed us to detect hexanal down to 1 fM
Laura Purriños et al.
Meat science, 93(2), 344-350 (2012-10-30)
The volatile profile of dry-cured "lacón" that has been inoculated with three different yeasts were determined and compared with a non-inoculated dry-cured "lacón". Yeasts (Debaryomyces hansenii, Candida deformans and Candida zeylanoides) that were used as starter cultures in the present
F Masotti et al.
Journal of dairy science, 95(9), 4760-4767 (2012-08-25)
This study investigated the changes in chemical and microbiological properties of fresh goat milk cheese stored in an open deck refrigerated display cabinet (6 ± 2°C) or in a dark cold room (4 ± 1°C). The effects of partial-vacuum packaging
Laura Purriños et al.
Meat science, 92(4), 627-634 (2012-07-04)
The effect of the length of salting time on volatile compounds throughout the manufacture of dry-cured "lacón" was studied using a purge-and-trap extraction and GC/MS analysis. For this study, six treatments of "lacón" were salted with different amounts of salt
Brian Farneti et al.
Journal of food science, 77(5), C551-C559 (2012-04-19)
The availability of rapid and accurate methods to assess fruit flavor is of utmost importance to support quality control especially in the breeding phase. Breeders need more information and analytical tools to facilitate selection for complex multigenic traits such as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service