Skip to Content
Merck
All Photos(1)

Key Documents

W254002

Sigma-Aldrich

Heptanal

≥95%, FCC, FG

Synonym(s):

Heptaldehyde, 1-Heptanal, Aldehyde C7, Enanthaldehyde, Oenanthaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5CHO
CAS Number:
Molecular Weight:
114.19
FEMA Number:
2540
Beilstein:
1560236
EC Number:
Council of Europe no.:
117
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
5.031
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

300
400

grade

FG
Fragrance grade
Halal

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥95%

refractive index

n20/D 1.413 (lit.)

bp

153 °C (lit.)

mp

−43 °C (lit.)

density

0.817 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

fatty; fresh; green

SMILES string

CCCCCCC=O

InChI

1S/C7H14O/c1-2-3-4-5-6-7-8/h7H,2-6H2,1H3

InChI key

FXHGMKSSBGDXIY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

95.0 °F - closed cup

Flash Point(C)

35 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Romina B Barrozo et al.
The European journal of neuroscience, 33(10), 1841-1850 (2011-04-15)
Innate behaviours in animals can be influenced by several factors, such as the environment, experience, or physiological status. This behavioural plasticity originates from changes in the underlying neuronal substrate. A well-described form of plasticity is induced by mating. In both
Nina Deisig et al.
PloS one, 7(3), e33159-e33159 (2012-03-20)
Most animals rely on olfaction to find sexual partners, food or a habitat. The olfactory system faces the challenge of extracting meaningful information from a noisy odorous environment. In most moth species, males respond to sex pheromone emitted by females
Kevin Huvaere et al.
Journal of agricultural and food chemistry, 59(16), 8718-8723 (2011-07-05)
The effect of two different antioxidants, EDTA and green tea extract (GTE), used individually or in combination, on the light-induced oxidation of reduced fat soft cheeses (0.2 and 6% fat) was investigated. In samples with 0.2% fat, lipid hydroperoxides as
Ning Li et al.
Analytical biochemistry, 342(2), 318-326 (2005-05-26)
In this work, we developed a new approach to the analysis of the lung cancer biomarkers, hexanal and heptanal in human blood that was based on headspace single-drop microextraction (HS-SDME) with droplet derivatization, followed by gas chromatography-mass spectrometry (GC-MS). Aldehydes
Rossella Guadagni et al.
Analytica chimica acta, 701(1), 29-36 (2011-07-19)
Hexanal and heptanal are endogenous aldehydes coming from membrane lipid oxidation, found in lung cancer patients' blood, and suggested as lung tumor biomarkers. Here the urinary matrix was investigated instead of blood and the difficulties related to the determination of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service