Merck
  • Simultaneous Enantioseparation of Aldohexoses and Aldopentoses Derivatized With L-Tryptophanamide by Reversed Phase HPLC Using Butylboronic Acid as a Complexation Reagent of Monosaccharides.
Simultaneous Enantioseparation of Aldohexoses and Aldopentoses Derivatized With L-Tryptophanamide by Reversed Phase HPLC Using Butylboronic Acid as a Complexation Reagent of Monosaccharides.
Chirality (2015-05-23)
Mirei Shou, Hiroyuki Terashima, Sen-Ichi Aizawa, Atsushi Taga, Atsushi Yamamoto, Shuji Kodama
ABSTRACT
Three aldohexoses, glucose, galactose, and mannose, and three aldopentoses, arabinose, xylose, and ribose, were derivatized with L-tryptophanamide (L-TrpNH2 ) under alkaline conditions. Using a basic mobile phase (pH 9.2), the three aldohexoses or the three aldopentoses were simultaneously enantioseparated, respectively, but all the six monosaccharides could not be simultaneously enantioseparated. A large amount of nonreacted L-TrpNH2 was detected after the derivatized monosaccharides. In order to widen the separation window, a large portion of nonreacted L-TrpNH2 could be eliminated by liquid-liquid extraction with ethylacetate, and elution order of the derivatized monosaccharides and nonreacted L-TrpNH2 was found to be reversed using a neutral mobile phase. All of the six monosaccharides were simultaneously enantioseparated by reversed phase high-performance liquid chromatography (HPLC) using InertSustainSwift C18 column (4.6 mm i.d. × 150 mm) and a mobile phase containing 180 mM phosphate buffer (pH 7.6), 1.5 mM butylboronic acid, and 5% acetonitrile at 40 °C. Nomenclature of D and L for monosaccharides is based on the configurations of the asymmetric C4 center for aldopentoses and C5 center for aldohexoses. It was found that the enantiomer elution order of these six monosaccharides and fucose in the proposed method conformed to be the absolute configuration of the C2 center.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Cyclohexylboronic acid, ≥95%
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
L-(−)-Fucose, ≥98% (GC)
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate
Sigma-Aldrich
Ethyl acetate