Skip to Content
Merck
All Photos(1)

Key Documents

W530574

Sigma-Aldrich

Hydrochloric acid solution

32 wt. % in H2O, FCC

Synonym(s):

Muriatic acid, Hydrogen chloride solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HCl
CAS Number:
Molecular Weight:
36.46
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
NACRES:
NA.21
grade:
Kosher
biological source:
synthetic
food allergen:
no known allergens
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

grade

Kosher

reg. compliance

FCC

vapor pressure

190 mmHg ( 25 °C)

description

20 degrees baumé

form

liquid

concentration

32 wt. % in H2O

bp

100 °C (lit.)

density

1.161 g/mL at 25 °C

anion traces

sulfate (SO42-): ≤0.5%

cation traces

As: ≤3 ppm
Cd: ≤1 ppm
Fe: ≤5 ppm
Hg: ≤0.1 ppm
Pb: ≤1 ppm

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

faint

SMILES string

Cl

InChI

1S/ClH/h1H

InChI key

VEXZGXHMUGYJMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Hydrochloric acid solution (32 wt. % in H2O, FCC) is used in the food industry as pH adjusters [1] and food additives (2).

Application

Hydrochloric acid solution is used as a pH adjuster:
  • In the production of high fructose corn syrup (HFCS) during the process of converting corn starch to fructose. [1]
  • In the production of drinking water, beverages and foods. (2)
Hydrochloric acid solution is also used as a processing aid in the production of food additives like fructose, citric acid, and hydrolyzed vegetable protein. (2)

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High fructose corn syrup: production, uses and public health concerns
Parker K, et al.
Biotechnology and Molecular Biology Reviews, 5(5), 71-78 (2010)
Douglas M McMillan et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 40(7), 1804-1812 (2015-01-30)
CYP2D metabolically activates codeine to morphine, which is required for codeine analgesia. Permeability across the blood-brain barrier, and active efflux, suggests that initial morphine in the brain after codeine is due to brain CYP2D metabolism. Human CYP2D is higher in
Xuan Zhang et al.
Nanomedicine : nanotechnology, biology, and medicine, 11(6), 1365-1375 (2015-04-19)
Intratumoral injection of ultra-small gold nanoparticles (AuNPs) conjugated to doxorubicin (Au-Dox) is effective against both murine B16 and human SK-MEL-28 tumors in mice. Au-Dox suppresses growth of B16 tumors in immunocompetent mice by >70% for at least 19 days. In
Yosuke Senju et al.
Journal of cell science, 128(15), 2766-2780 (2015-06-21)
PACSIN2, a membrane-sculpting BAR domain protein, localizes to caveolae. Here, we found that protein kinase C (PKC) phosphorylates PACSIN2 at serine 313, thereby decreasing its membrane binding and tubulation capacities. Concomitantly, phosphorylation decreased the time span for which caveolae could
Rebecca J Ford et al.
The Biochemical journal, 468(1), 125-132 (2015-03-06)
Metformin is the mainstay therapy for type 2 diabetes (T2D) and many patients also take salicylate-based drugs [i.e., aspirin (ASA)] for cardioprotection. Metformin and salicylate both increase AMP-activated protein kinase (AMPK) activity but by distinct mechanisms, with metformin altering cellular

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service