Skip to Content
Merck
All Photos(3)

Key Documents

270989

Sigma-Aldrich

Ethyl acetate

Ethyl acetate
1 of 2 reviewers received a sample product or took part in a promotion

anhydrous, 99.8%

Synonym(s):

EtOAc

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOC2H5
CAS Number:
Molecular Weight:
88.11
Beilstein:
506104
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.21
grade:
anhydrous
bp:
76.5-77.5 °C (lit.)
vapor pressure:
73 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

anhydrous

Quality Level

vapor density

3 (20 °C, vs air)

vapor pressure

73 mmHg ( 20 °C)

product line

SAFC Hitech®

Assay

99.8%

form

liquid

autoignition temp.

801 °F

expl. lim.

2.2-11.5 %, 38 °F

impurities

<0.005% water

evapn. residue

<0.0005%

refractive index

n20/D 1.3720 (lit.)

bp

76.5-77.5 °C (lit.)

mp

−84 °C (lit.)

solubility

water: soluble

density

0.902 g/mL at 25 °C (lit.)

format

neat

SMILES string

CCOC(C)=O

InChI

1S/C4H8O2/c1-3-6-4(2)5/h3H2,1-2H3

InChI key

XEKOWRVHYACXOJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl acetate is an effective alternate solvent of diethyl ether, employed for the concentration of eggs, larvae, and cysts in fecal specimens during the Formalin-ether sedimentation technique.[1] Acetaldehyde is reaction intermediate formed during the oxidative combustion of ethanol and ethyl acetate in the presence of copper oxide embedded on Ce-doped titania surface.[2]

Application

Ethyl acetate may be used as solvent for the synthesis of γ-benzyl-L-glutamic acid N-carboxy anhydride (BLG-NCA).[3]

Legal Information

SAFC Hitech is a registered trademark of Sigma-Aldrich Co. LLC

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

24.8 °F - closed cup

Flash Point(C)

-4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fernando E Maturi et al.
Materials (Basel, Switzerland), 12(6) (2019-03-25)
Mesoporous silica nanoparticles prepared by organic template-driven synthesis have been successfully explored as carriers of the drug-derivate green luminescent complex of terbium (III) with the nonsteroidal anti-inflammatory drug ketoprofen. The terbium (III) complex was synthesized by reacting ketoprofen sodium salt
Andreas F Thünemann et al.
Langmuir : the ACS journal of surfaces and colloids, 22(5), 2351-2357 (2006-02-24)
Superparamagnetic iron oxide particles (SPIO) of maghemite were prepared in aqueous solution and subsequently stabilized with polymers in two layer-by-layer deposition steps. The first layer around the maghemite core is formed by poly(ethylene imine) (PEI), and the second one is
Complete oxidation of CO, ethanol, and ethyl acetate over copper oxide supported on titania and ceria modified titania.
Larsson PO and Andersson A.
J. Catal., 179(1), 72-89 (1998)
Izabela N Faria Gomes et al.
Molecules (Basel, Switzerland), 24(21) (2019-11-07)
Plant-based compounds are an option to explore and perhaps overcome the limitations of current antitumor treatments. Annona coriacea Mart. is a plant with a broad spectrum of biological activities, but its antitumor activity is still unclear. The purpose of our
K H Young et al.
Journal of clinical microbiology, 10(6), 852-853 (1979-12-01)
Ethyl acetate appears to be a satisfactory subsitute solvent for diethyl ether in the Formalin-ether sedimentation technique. In comparative studies, concentration of organisms with ethyl acetate was equal to or greater than that with diethyl ether. No distortion or alteration

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

1 of 2 reviewers received a sample product or took part in a promotion

Active Filters

  1. 2 Ratings-Only Reviews

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service