Merck
  • Male fertility and apoptosis in normal spermatogenesis are regulated by vacuolar-ATPase isoform a2.
Male fertility and apoptosis in normal spermatogenesis are regulated by vacuolar-ATPase isoform a2.
Journal of reproductive immunology (2015-08-01)
Mukesh K Jaiswal, Varkha Agrawal, Gajendra K Katara, Sahithi Pamarthy, Arpita Kulshrestha, Gerard Chaouat, Alice Gilman-Sachs, Kenneth D Beaman
ABSTRACT
The a2 isoform of vacuolar-ATPase (ATP6V0A2, referred to as a2V) is required for normal spermatogenesis and maturation of sperm. Treatment of male mice with anti-a2V disturbs the testicular cytokine/chemokine balance and leads to severe deficiencies of spermatogenesis. The aim of the present study was to investigate the role of a2V in male fertility and in the regulation of apoptotic pathways required for normal spermatogenesis in mice. To study the role of a2V single dose of anti-a2V monoclonal antibody or mouse IgG isotype (3μg/animal) was injected i.p. into males on alternate days for 10 days. The expression of sperm maturation-related molecules and pro-apoptotic molecules was measured by real-time PCR or immunohistochemistry in control and anti-a2V-treated testes. The caspase levels and their activity were measured by western blot and fluorometry. We found that the expression of the sperm maturation-related molecules SPAM1, ADAM1, and ADAM2 was significantly decreased in testes from anti-a2V-treated males. The expression of pro-apoptotic molecules (Bax, p53, and p21) and molecules involved in the intrinsic pathway of apoptosis (caspase-9, caspase-3, and PARP), which are crucial for normal spermatogenesis was significantly reduced in testes from anti-a2V-treated males compared with the control. The total ATP level was significantly lower in anti-a2V-treated testes. The data provide novel evidence showing that a2V can regulate the apoptotic pathways, an essential testicular feature, and is necessary for efficient spermatogenesis.
Sigma-Aldrich
Silica, nanopowder, 99.8% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Silica, mesostructured, MSU-F (cellular foam)
Pricing and availability is not currently available.
Sigma-Aldrich
1-Naphthyl phosphate monosodium salt monohydrate, ≥98% (titration), powder
Pricing and availability is not currently available.
Sigma-Aldrich
Silica, fumed, powder
Pricing and availability is not currently available.
Sigma-Aldrich
Silica, fumed, powder, 0.2-0.3 μm avg. part. size (aggregate)
Pricing and availability is not currently available.
Sigma-Aldrich
Silica
Pricing and availability is not currently available.
Sigma-Aldrich
Silicon dioxide, granular, ≥99.9%
Pricing and availability is not currently available.
Sigma-Aldrich
Silicon dioxide, acid washed
Pricing and availability is not currently available.
Sigma-Aldrich
Silicon dioxide, washed and calcined, analytical reagent
Pricing and availability is not currently available.
Supelco
Silica, 99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
LUDOX® SM colloidal silica, 30 wt. % suspension in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Silicon dioxide, fused (granular), 1-3 mm, 99.9% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Silicon dioxide, −325 mesh, 99.5% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
LUDOX® LS colloidal silica, 30 wt. % suspension in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
LUDOX® HS-30 colloidal silica, 30 wt. % suspension in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
LUDOX® TM-40 colloidal silica, 40 wt. % suspension in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
LUDOX® CL colloidal silica, 30 wt. % suspension in H2O
Pricing and availability is not currently available.
Supelco
Glass spheres
Pricing and availability is not currently available.
Sigma-Aldrich
Silicon dioxide, acid washed and calcined, Analytical Reagent
Pricing and availability is not currently available.
Sigma-Aldrich
Silica, nanoparticles, mesoporous, 200 nm particle size, pore size 4 nm
Pricing and availability is not currently available.
Sigma-Aldrich
Silicon dioxide, single crystal substrate, optical grade, 99.99% trace metals basis, L × W × thickness 10 mm × 10 mm × 0.5 mm
Pricing and availability is not currently available.
Sigma-Aldrich
Silicon dioxide, nanopowder (spherical, porous), 5-20 nm particle size (TEM), 99.5% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Silica, mesostructured, SBA-15, 99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Silica, mesostructured, MCM-41 type (hexagonal)
Pricing and availability is not currently available.
Sigma-Aldrich
Silicon dioxide, nanopowder, 10-20 nm particle size (BET), 99.5% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Silicon dioxide, ~99%, 0.5-10 μm (approx. 80% between 1-5 μm)
Pricing and availability is not currently available.
Sigma-Aldrich
Silicon dioxide, purum p.a., acid purified
Pricing and availability is not currently available.
Sigma-Aldrich
Silicon dioxide, purum p.a., acid purified, sand
Pricing and availability is not currently available.
Sigma-Aldrich
LUDOX® AM colloidal silica, 30 wt. % suspension in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
LUDOX® CL-X colloidal silica, 45 wt. % suspension in H2O
Pricing and availability is not currently available.