Skip to Content
Merck
All Photos(3)

Documents

342831

Sigma-Aldrich

Silicon dioxide

fused (granular), 4-20 mesh, 99.9% trace metals basis

Synonym(s):

Cristobalite, Quartz, Sand, Silica

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SiO2
CAS Number:
Molecular Weight:
60.08
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9% trace metals basis

form

fused (granular)

refractive index

n20/D 1.544 (lit.)

particle size

4-20 mesh

mp

1610 °C (lit.)

density

2.6 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

SiO2 is majorly used as a substrate material with excellent thermo-mechanical properties, which can be used in a variety of applications which include: vapor deposition, phase deposition, atomic force microscopy probes (AFM), spin coating, electronic based devices.

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The effect of calcination temperature on the surface microstructure and photocatalytic activity of TiO2 thin films prepared by liquid phase deposition.
Yu J, et al.
The Journal of Physical Chemistry B, 107(50), 13871-13879 (2003)
Exciton diffusion measurements in poly (3-hexylthiophene).
Shaw PE, et al.
Advanced Materials, 20(18), 3516-3520 (2008)
Electrical and optical properties of ZnO transparent conducting films by the sol-gel method.
Lee J, et al.
Journal of Crystal Growth, 247(1-2), 119-125 (2003)
Oxygen-aided synthesis of polycrystalline graphene on silicon dioxide substrates.
Chen J, et al.
Journal of the American Chemical Society, 133(44), 17548-17551 (2011)
Measurement of the adhesion force between carbon nanotubes and a silicon dioxide substrate.
Whittaker JD, et al.
Nano Letters, 6(5), 953-957 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service