Skip to Content
Merck
All Photos(1)

Key Documents

440345

Supelco

Glass spheres

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
O2Si
CAS Number:
Molecular Weight:
60.08
EC Number:
MDL number:
UNSPSC Code:
23201100
PubChem Substance ID:
NACRES:
SB.52

grade

high-purity grade

form

powder

technique(s)

LPLC: suitable

surface area

125 m2/g

matrix

Silica

matrix active group

silica

mean particle size

9-13 μm

density

1.1 g/mL at 25 °C (lit.)

suitability

suitable for acidity or alkalinity (alkalinity < 0.5 meq/g ASTM D 3100)

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Silica is a naturally occurring material in minerals, flint and in some plants in crystalline phase. Silica used in industries is in synthetic form. Surface area, pore volume, pore size and particle size are independently controllable to some extent. The crystalline silica may be classified based on atmospheric pressure as:
a. Quartz = 1143K
b. Tridymite 1143 - 1743K
c. Cristobalite = 1743, over 1973K it forms amorphous vitreous silica glass.

Application

Low density filler for coatings and plastics.
Silica glass spheres are low density filler for coatings and plastics. Silica was used in synthesis of silicalite. Silica was also used to assist the catalytic growth of oxide and nitride nanowires.

Features and Benefits

Light weight, crush-resistant filler. Imparts dimensional stability, reduces warpage, reduces plastic melt viscosity and provides multidirectional orientation in reinforced composites.

Physical form

Hollow, spherical non-porous glass beads.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Norma A Sánchez-Flores et al.
Journal of colloid and interface science, 323(2), 359-364 (2008-05-27)
Well crystallized silicalite-1 has been obtained from three sources of amorphous silica, namely, rice hull ashes, commercial Davisil, and a fume silica from Aldrich. The silicas were first dissolved in glycerol according to a recently described reaction. This reaction transforms
The Heat Capacities of Quartz, Cristobalite and Tridymite at Low Temperatures
Anderson, C. Travis.
Journal of the American Chemical Society, 58, 568-570 (1936)
Silica-assisted catalytic growth of oxide and nitride nanowires.
Tang, C. C., et al.
Chemical Physics Letters, 333, 12-15 (2001)
Yijun Yao et al.
Journal of hazardous materials, 298, 221-231 (2015-06-10)
In 2002, U.S. EPA proposed a general buffer zone of approximately 100 feet (30 m) laterally to determine which buildings to include in vapor intrusion (VI) investigations. However, this screening distance can be threatened by factors such as extensive surface
Keren B Turton et al.
PloS one, 10(5), e0127243-e0127243 (2015-05-20)
Signal transducer and activator of transcription 3 (STAT3) is a key mediator of leukocyte differentiation and proliferation. The 3' end of STAT3 transcripts is subject to two alternative splicing events. One results in either full-length STAT3α or in STAT3β, which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service