Skip to Content
Merck
All Photos(1)

Key Documents

420808

Sigma-Aldrich

LUDOX® LS colloidal silica

30 wt. % suspension in H2O

Synonym(s):

Silica preparation

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SiO2
CAS Number:
Molecular Weight:
60.08
MDL number:
UNSPSC Code:
12352309
PubChem Substance ID:
NACRES:
NA.22

description

colloidal

form

viscous liquid

concentration

30 wt. % suspension in H2O

surface area

~215 m2/g

pH

8.2

density

1.21 g/mL at 25 °C

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

sodium stabilizing counterion

Legal Information

LUDOX is a registered trademark of W.R. Grace & Co.-Conn.
LUDOX is a registered trademark of W.R. Grace & Co.-Conn.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David M Brown et al.
Toxicology letters, 224(1), 147-156 (2014-01-28)
We investigated the effects of silica particles and nanoparticles (NPs) (50 nm and 200 nm) with a neutral and positively charged surface when dispersed in saline, bovine serum albumin (BSA) or lung lining fluid (LLF) 24 h post instillation into
Richard Hayes et al.
Journal of chromatography. A, 1357, 36-52 (2014-05-27)
The challenges in HPLC are fast and efficient separation for a wide range of samples. Fast separation often results in very high operating pressure, which places a huge burden on HPLC instrumentation. In recent years, core-shell silica microspheres (with a
Fabrice Gritti et al.
Journal of chromatography. A, 1342, 24-29 (2014-04-17)
The advantages of using columns packed with stationary phases having a composition gradient so that retention factors increase toward the outlet and eluting them with an isocratic mobile phase may provide increased peak resolution in liquid chromatography. This approach is
Alexandra I Lupulescu et al.
Science (New York, N.Y.), 344(6185), 729-732 (2014-05-17)
The growth mechanism of silicalite-1 (MFI zeolite) is juxtaposed between classical models that postulate silica molecules as primary growth units and nonclassical pathways based on the aggregation of metastable silica nanoparticle precursors. Although experimental evidence gathered over the past two
Graham J Edgar et al.
Nature, 506(7487), 216-220 (2014-02-07)
In line with global targets agreed under the Convention on Biological Diversity, the number of marine protected areas (MPAs) is increasing rapidly, yet socio-economic benefits generated by MPAs remain difficult to predict and under debate. MPAs often fail to reach

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service