Skip to Content
Merck
All Photos(1)

Key Documents

381276

Supelco

Silica Inorganic Sorbent

120-140 mesh, 99.8%

Synonym(s):

Silica, fumed, Silica, Silicic anhydride, Silicon dioxide, Silicon dioxide amorphous

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SiO2
CAS Number:
Molecular Weight:
60.08
MDL number:
UNSPSC Code:
23201100
PubChem Substance ID:
NACRES:
SB.52

Product Name

Silica, 99.8%

grade

standard

Quality Level

Assay

99.8%

form

powder

technique(s)

LPLC: suitable

surface area

175-225 m2/g

matrix

Silica

matrix active group

silica

particle size

0.011 μm

pore size

60 Å mean pore size

separation technique

hydrophilic interaction (HILIC)

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Silica is a naturally occurring material in minerals, flint and in some plants in crystalline phase. Silica commonly used in industries is in synthetic form. Surface area, pore volume, pore size and particle size are independently controllable to some extent. The crystalline silica may be classified based on atmospheric pressure as:
a. Quartz: = 1143K
b. Tridymite: 1143 - 1743K
c. Cristobalite: = 1743, over 1973K it forms amorphous vitreous silica glass.

Application

Silica was used in synthesis of silicalite. Silica was used to assist the catalytic growth of oxide and nitride nanowires.

Features and Benefits

Has interesting thickening and thixotropic properties, and an enormous external surface area.

Other Notes

Particle sizes are for primary particles, which form branched, chain-like aggregates a few tenths of a micron long.
May contain adsorbed H2O and CO2 which is removable by calcining at >900°C

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohadese Farhangian et al.
Industrial health, 59(6), 415-426 (2021-10-01)
The aim of the present study was to first fabricate an electrospun PVC nanofiber web and then assess its applicability in sampling and measuring the concentration of airborne crystalline silica by comparing analysis results with a commercial PVC membrane filter
Silica-assisted catalytic growth of oxide and nitride nanowires.
Tang, C. C., et al.
Chemical Physics Letters, 333, 12-15 (2001)
Norma A Sánchez-Flores et al.
Journal of colloid and interface science, 323(2), 359-364 (2008-05-27)
Well crystallized silicalite-1 has been obtained from three sources of amorphous silica, namely, rice hull ashes, commercial Davisil, and a fume silica from Aldrich. The silicas were first dissolved in glycerol according to a recently described reaction. This reaction transforms
The Heat Capacities of Quartz, Cristobalite and Tridymite at Low Temperatures
Anderson, C. Travis.
Journal of the American Chemical Society, 58, 568-570 (1936)
Srivalleesha Mallidi et al.
Nano letters, 9(8), 2825-2831 (2009-07-04)
Gold nanoparticles targeting epidermal growth factor receptor via antibody conjugation undergo molecular specific aggregation when they bind to receptors on cell surfaces, leading to a red shift in their plasmon resonance frequency. Capitalizing on this effect, we demonstrate the efficacy

Articles

Operation principle and market dominance of single crystalline silicon solar cells.

Operation principle and market dominance of single crystalline silicon solar cells.

Operation principle and market dominance of single crystalline silicon solar cells.

Operation principle and market dominance of single crystalline silicon solar cells.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service