Skip to Content
Merck
All Photos(1)

Documents

N7510

Sigma-Aldrich

Nicardipine hydrochloride

powder, ≥98%

Synonym(s):

1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic acid methyl 2-[methyl(phenylmethyl)amino]ethyl ester hydrochloride, YC-93

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H29N3O6 · HCl
CAS Number:
Molecular Weight:
515.99
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98%

form

powder

color

yellow

solubility

DMSO: ~1 mg/mL
0.1 M NaOH: insoluble
H2O: slightly soluble
methanol: soluble

originator

EKR Therapeutics

storage temp.

2-8°C

SMILES string

Cl[H].COC(=O)C1=C(C)NC(C)=C(C1c2cccc(c2)[N+]([O-])=O)C(=O)OCCN(C)Cc3ccccc3

InChI

1S/C26H29N3O6.ClH/c1-17-22(25(30)34-4)24(20-11-8-12-21(15-20)29(32)33)23(18(2)27-17)26(31)35-14-13-28(3)16-19-9-6-5-7-10-19;/h5-12,15,24,27H,13-14,16H2,1-4H3;1H

InChI key

AIKVCUNQWYTVTO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Blocks L-type voltage-dependent calcium channels; antihypertensive.

Features and Benefits

This compound was developed by EKR Therapeutics. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T J Hibberd et al.
Neurogastroenterology and motility : the official journal of the European Gastrointestinal Motility Society, 29(10), 1-12 (2017-04-19)
Relatively little is known about the electrical rhythmicity of the whole colon, where long neural pathways are preserved. Smooth muscle electrical activity was recorded extracellularly from the serosa of isolated flat-sheet preparations consisting of the whole mouse colon (n=31). Two
GoEun Han et al.
Frontiers in neuroscience, 16, 969712-969712 (2022-11-08)
Non-rapid eye movement (NREM) sleep is tightly homeostatically regulated and essential for survival. In the electroencephalogram (EEG), oscillations in the delta (0.5-4 Hz) range are prominent during NREM sleep. These delta oscillations are, to date, the best indicator for homeostatic
S Thellung et al.
Neurobiology of disease, 7(4), 299-309 (2000-08-31)
Prion diseases are neurodegenerative pathologies characterized by the accumulation, in the brain, of altered forms of the prion protein (PrP), named PrP(Sc). A synthetic peptide homologous to residues 106-126 of PrP (PrP106-126) was reported to maintain the neurodegenerative characteristics of
Prasanna P K M Gamage et al.
Journal of cellular and molecular medicine, 22(10), 5160-5164 (2018-07-27)
Ageing is associated with impaired neuromuscular function of the terminal gastrointestinal (GI) tract, which can result in chronic constipation, faecal impaction and incontinence. Interstitial cells of cajal (ICC) play an important role in regulation of intestinal smooth muscle contraction. However
Avril Coghlan et al.
PLoS neglected tropical diseases, 17(9), e0011205-e0011205 (2023-09-05)
Hundreds of millions of people worldwide are infected with the whipworm Trichuris trichiura. Novel treatments are urgently needed as current drugs, such as albendazole, have relatively low efficacy. We have investigated whether drugs approved for other human diseases could be

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service