Skip to Content
Merck

T3000000

Tyrothricin

European Pharmacopoeia (EP) Reference Standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

tyrothricin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

N21[C@@H](CCC2)C(=O)N[C@H](C(=O)N[C@@H](C(=O)[C@@H](NC(=O)[C@@H](N[C@H](C(=O)N[C@H](C(=O)N[C@H](C(=O)N[C@H](C(=O)N[C@@H](C1=O)Cc6ccccc6)CC(C)C)CCCN)C(C)C)Cc5ccc(cc5)O)CCC(=O)O)CC(=O)N)Cc4ccccc4)Cc3ccccc3

InChI

1S/C65H85N11O13/c1-38(2)32-49-60(84)74-52(36-42-20-12-7-13-21-42)65(89)76-31-15-23-53(76)63(87)73-51(34-41-18-10-6-11-19-41)61(85)70-47(33-40-16-8-5-9-17-40)57(81)48(37-54(67)78)71-59(83)46(28-29-55(79)80)68-50(35-43-24-26-44(77)27-25-43)62(86)75-56(39(3)4)64(88)69-45(22-14-30-66)58(82)72-49/h5-13,16-21,24-27,38-39,45-53,56,68,77H,14-15,22-23,28-37,66H2,1-4H3,(H2,67,78)(H,69,88)(H,70,85)(H,71,83)(H,72,82)(H,73,87)(H,74,84)(H,75,86)(H,79,80)/t45-,46-,47+,48-,49-,50-,51-,52+,53-,56-/m0/s1

InChI key

NLJVXZFCYKWXLH-DXTIXLATSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Tyrothricin EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.


Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



W Wigger-Alberti et al.
Skin pharmacology and physiology, 26(1), 52-56 (2012-11-28)
Topical preparations are a common treatment for superficial acute wounds, which at the least do not interfere with healing and ideally result in enhanced wound healing irrespective of microbial colonization. To examine the effects of a topical antimicrobial gel and
Sidharth Chopra et al.
The Journal of antimicrobial chemotherapy, 65(12), 2598-2601 (2010-09-24)
The rising occurrence of drug-resistant pathogens accentuates the need to identify novel antibiotics. We wanted to identify new scaffolds for drug discovery by repurposing FDA-approved drugs against Acinetobacter baumannii, an emerging Gram-negative nosocomial drug-resistant pathogen. In this study, we screened
[Copper and manganese gluconate in the treatment of breast fissures during breast feeding].
J Barrat et al.
Revue francaise de gynecologie et d'obstetrique, 87(1), 49-51 (1992-01-01)



Global Trade Item Number

SKUGTIN
T300000004061837343605