Skip to Content
Merck
All Photos(1)

Documents

CDS023093

Sigma-Aldrich

Nilotinib

Synonym(s):

4-Methyl-3-[[4-(3-pyridinyl)-2-pyrimidinyl]amino]-N-[5-(4-methyl-1H-imidazol-1-yl)-3-(trifluoromethyl)phenyl]benzamide, 4-Methyl-N-[3-(4-methyl-1H-imidazol-1-yl)-5-(trifluoromethyl)phenyl]-3-[[4-(3-pyridinyl)-2-pyrimidinyl]amino]benzamide, 4-Methyl-N-[3-(4-methylimidazol-1-yl)-5-trifluoromethylphenyl]-3-[[4-(pyridin-3-yl)pyrimidin-2-yl]amino]benzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H22F3N7O
CAS Number:
Molecular Weight:
529.52
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

description

AldrichCPR

form

solid

storage temp.

-10 to -25°C

SMILES string

CC1=CN(C2=CC(NC(C3=CC=C(C(NC4=NC=CC(C5=CC=CN=C5)=N4)=C3)C)=O)=CC(C(F)(F)F)=C2)C=N1

InChI

1S/C28H22F3N7O/c1-17-5-6-19(10-25(17)37-27-33-9-7-24(36-27)20-4-3-8-32-14-20)26(39)35-22-11-21(28(29,30)31)12-23(13-22)38-15-18(2)34-16-38/h3-16H,1-2H3,(H,35,39)(H,33,36,37)

InChI key

HHZIURLSWUIHRB-UHFFFAOYSA-N

Gene Information

human ... ABL1(25)

General description

Nilotinib is an aminopyrimidine-derivative that can be used as a selective inhibitor of tyrosine kinase receptors. It also has a role as an antineoplastic agent and an anticoronaviral agent.

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY, (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE, OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY, WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - STOT RE 1

Target Organs

Liver,Kidney,gallbladder

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hans Gelderblom et al.
The Lancet. Oncology, 19(5), 639-648 (2018-03-25)
Pigmented villonodular synovitis (alternatively known as diffuse-type giant cell tumour) is a rare, locally aggressive tumour driven by a specific translocation resulting in the overexpression of colony-stimulating factor 1 (CSF1). CSF1 receptor (CSF1R) inhibitors (ie, tyrosine kinase inhibitors and antibodies)
A Hochhaus et al.
Leukemia, 31(7), 1525-1531 (2017-02-22)
The single-arm, phase 2 ENESTfreedom trial assessed the potential for treatment-free remission (TFR; i.e., the ability to maintain a molecular response after stopping therapy) following frontline nilotinib treatment. Patients with Philadelphia chromosome-positive chronic myeloid leukemia in chronic phase with MR
Fabienne Lamballe et al.
Oncotarget, 7(46), 74747-74767 (2016-10-13)
The cytoplasmic tyrosine kinase ABL exerts positive or negative effects in solid tumours according to the cellular context, thus functioning as a "switch modulator". The therapeutic effects of drugs targeting a set of signals encompassing ABL have been explored in
Main Chemotypes of SARS-CoV-2 Reproduction Inhibitors
Shiryaev VA and Klimochkin Yu N
Russian Journal of Organic chemistry, 57(5), 730-767 (2021)
Qinghui Yang et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 42(6), 2182-2193 (2017-08-17)
Cardiotoxicity is a predominant side-effect of nilotinib during chronic myeloid leukemia treatment. The underlying molecular mechanism remains unclear. The role of autophagy and mitochondrial signaling was investigated in nilotinib-treated cardiac H9C2 cells. Cytotoxicity was assessed using Cell Death Detection kit.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service