Skip to Content
Merck
All Photos(5)

Documents

G2253

Sigma-Aldrich

L-Glutamic acid γ-monohydroxamate

≥97% (TLC)

Synonym(s):

L-γ-Glutamylhydroxamic acid, L-5-N-Hydroxyglutamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10N2O4
CAS Number:
Molecular Weight:
162.14
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

product name

L-Glutamic acid γ-monohydroxamate,

Assay

≥97% (TLC)

form

powder

color

white to off-white

application(s)

detection

storage temp.

−20°C

SMILES string

NC(CCC(=O)NO)C(O)=O

InChI

1S/C5H10N2O4/c6-3(5(9)10)1-2-4(8)7-11/h3,11H,1-2,6H2,(H,7,8)(H,9,10)

InChI key

YVGZXTQJQNXIAU-UHFFFAOYSA-N

Application

L-Glutamic acid γ-monohydroxamate has been used as a standard to calculate transglutaminase (TGase) activity.

Biochem/physiol Actions

L-Glutamic acid γ-monohydroxamate [L-Glu(gamma)HXM] is used as a vanadium ligand which potentiates vanadiums metabolic activity. L-Glu(gamma)HXM is also used as a substrate for E. coli asparagine synthetase B and as an ATP-dependent irreversible inhibitor of Escherichia coli gamma-glutamylcysteine synthetase.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Seiler et al.
Neurochemical research, 15(3), 301-305 (1990-03-01)
The method for the assay of glutamine synthetase (GlnS) relies on the gamma-glutamyl transferase reaction, i.e. the formation of glutamyl-gamma-hydroxamate from glutamine and hydroxylamine, and the chromatographic separation of the reaction product from the reactants. The method is not only
X Huang et al.
The Journal of biological chemistry, 275(34), 26233-26240 (2000-08-22)
The x-ray crystal structure of the heterodimeric carbamoyl phosphate synthetase from Escherichia coli has identified an intermolecular tunnel that connects the glutamine binding site within the small amidotransferase subunit to the two phosphorylation sites within the large synthetase subunit. The
M Katoh et al.
Bioscience, biotechnology, and biochemistry, 62(7), 1455-1457 (1998-08-28)
Incubation of Escherichia coli gamma-glutamylcysteine synthetase with L-glutamic acid gamma-monohydroxamate and ATP caused slow but irreversible inhibition of the enzyme, and more than 90% activity was lost in three days. The enzyme was not inactivated when ATP was absent or
C J Lusty
FEBS letters, 314(2), 135-138 (1992-12-14)
E. coli carbamyl phosphate synthetase binds 0.2-0.4 mol equivalents of glutamine in an acid resistant form. The bound material is quantitatively released as glutamate by weak base hydrolysis and as a mixture of 12% glutamate, 10% gamma-glutamylhydroxamate, and 70% pyrrollidonecarboxylic
N Thomasset et al.
Anticancer research, 13(5A), 1393-1398 (1993-09-01)
We have previously shown that L-glutamic acid gamma-monohydroxamate (GAH) exhibits an antitumor activity, both in vitro and in vivo. In this report we explore the selective cytotoxicity of GAH in vitro by comparing the survival of tumor and normal cells.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service