Skip to Content
Merck
All Photos(3)

Key Documents

G8415

Sigma-Aldrich

L-Glutamic acid

98.5-100.5%, suitable for cell culture, non-animal source, meets EP testing specifications

Synonym(s):

(S)-2-Aminopentanedioic acid, Glu

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCH2CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
147.13
Beilstein:
1723801
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26
Pricing and availability is not currently available.

Product Name

L-Glutamic acid, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-100.5%

biological source

non-animal source

Quality Level

Agency

meets EP testing specifications

Assay

98.5-100.5%

form

powder

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

color

white

mp

205 °C (dec.) (lit.)

solubility

1 M HCl: 100 mg/mL

density

1.54 g/cm3 at 20 °C

anion traces

chloride (Cl-): ≤200 ppm
sulfate (SO42-): ≤200 ppm

cation traces

As: ≤1 ppm, passes test
Fe: ≤10 ppm, passes test
NH4+: ≤200 ppm, passes test

application(s)

pharmaceutical (small molecule)

SMILES string

N[C@@H](CCC(O)=O)C(O)=O

InChI

1S/C5H9NO4/c6-3(5(9)10)1-2-4(7)8/h3H,1-2,6H2,(H,7,8)(H,9,10)/t3-/m0/s1

InChI key

WHUUTDBJXJRKMK-VKHMYHEASA-N

Looking for similar products? Visit Product Comparison Guide

Application

L-Glutamic acid has been used as a supplement in for cell culture media to culture hippocampal neurons and hT-PA (human tissue plasminogen activation) producing cell line.[1][2][3]

Biochem/physiol Actions

Glutamic acid, or glutamate, the salt form of glutamic acid, functions as a neurotransmitter and also serves as a precursor to other neurotransmitters such as γ-aminobutyric acid.[4] Glutamic acid also plays a key role in many metabolic pathways, that controls growth, reproduction, maintenance and immunity.[5] It is converted to α-ketoglutarate, a key component of the TCA (tricarboxylic acid) cycle, and a precursor for the biosynthesis of nucleic acids and certain amino acids.[5][6] In cells, glutamine is converted to glutamate by the enzyme glutaminase.[6] Glutamine serves as a source of energy for rapidly dividing cells comprising lymphocytes, enterocytes, macrophages and tumors. Glutamine mediates protein turnover via cellular mTOR (mammalian target of rapamycin) signaling. It is also known to be associated with the inhibition of apoptosis.[5]
An excitatory amino acid neurotransmitter that is an agonist at all subtypes of glutamate receptors (metabotropic, kainate, NMDA, and AMPA).

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

The Biology of Cancer: Metabolic Reprogramming Fuels Cell Growth and Proliferation
Cell Metabolism, 11-20 (2008)
Geometric effect of cell adhesive polygonal micropatterns on neuritogenesis and axon
guidance
Min Jee Jang and Yoonkey Nam
Journal of Neural Engineering (2012)
Differential Effect of Culture Temperature and Specific Growth Rate on CHO Cell Behavior in Chemostat Culture
Vergara M9
PLoS ONE, 9(4) (2001)
Aqueous micro-contact printing of cell-adhesive biomolecules for patterning neuronal cell cultures.
Jang MJ, et al.
BioChip Journal, 6(2), 107-113 (2012)
Glutamic Acid, Twenty Years Later
S. Garattini
The Journal of Nutrition, 130(4), 901S-909S (2000)

Articles

Sigma article discusses tumor cell metabolic pathways, focusing on aerobic glycolysis and mitochondrial activity.

Sigma article discusses tumor cell metabolic pathways, focusing on aerobic glycolysis and mitochondrial activity.

Sigma article discusses tumor cell metabolic pathways, focusing on aerobic glycolysis and mitochondrial activity.

Sigma article discusses tumor cell metabolic pathways, focusing on aerobic glycolysis and mitochondrial activity.

Chromatograms

application for HPLC

Questions

1–3 of 3 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. I have to do a solution 147,13mg/ml Can I frooze aliquot of this solution?

    1 answer
    1. Unfortunately, information regarding the solution stability of Product G8415, L-Glutamic acid is not determined. It is recommended to dissolve the product and use freshly prepared dilutions.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service