Skip to Content
Merck
All Photos(10)

Documents

G4251

Sigma-Aldrich

L-Glutathione reduced

≥98.0%

Synonym(s):

Glutathione-SH, γ-L-Glutamyl-L-cysteinyl-glycine, GSH

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NCH(CO2H)CH2CH2CONHCH(CH2SH)CONHCH2CO2H
CAS Number:
Molecular Weight:
307.32
Beilstein:
1729812
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

product name

L-Glutathione reduced, ≥98.0%

Quality Level

Assay

≥98.0%

form

powder

color

white

mp

192-195 °C (dec.) (lit.)

solubility

water: 50 mg/mL, clear, colorless

application(s)

detection

functional group

amine
carboxylic acid
thiol

storage temp.

2-8°C

SMILES string

N[C@@H](CCC(=O)N[C@@H](CS)C(=O)NCC(O)=O)C(O)=O

InChI

1S/C10H17N3O6S/c11-5(10(18)19)1-2-7(14)13-6(4-20)9(17)12-3-8(15)16/h5-6,20H,1-4,11H2,(H,12,17)(H,13,14)(H,15,16)(H,18,19)/t5-,6-/m0/s1

InChI key

RWSXRVCMGQZWBV-WDSKDSINSA-N

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

γ-Glu-Cys-Gly

General description

Glutathione (GSH) is a ubiquitous molecule found in many cells and tissues. The three amino acids present in GSH are glycine, cysteine, and glutamic acid.

Application

L-Glutathione (GSH) reduced has been used in the elution buffer to elute GST (glutathione S-transferase)-fused proteins using glutathione-agarose beads. It has been used to prepare a standard curve for GSH analyses.
May be used at 5-10 mM to elute glutathione S-transferase (GST) from glutathione agarose.

Biochem/physiol Actions

Glutathione (GSH) forms conjugates with various metabolites and xenobiotics. It acts as an important co-factor for various enzymes. GSH is involved in many metabolic and signaling pathways. Glutathione functions as a thiol buffer for cellular proteins like thioredoxins and metallothioneins. GSH is also involved in the regeneration of antioxidants like tocopherols and ascorbate.
Endogenous antioxidant that plays a major role in reducing reactive oxygen species formed during cellular metabolism and the respiratory burst. Glutathione-S-transferase catalyzes the formation of glutathione thioethers with xenobiotics, leukotrienes, and other molecules that have an electrophilic center. Glutathione also forms disulfide bonds with cysteine residues in proteins. Via these mechanisms, it can have the paradoxical effect of reducing the efficacy of anti-cancer agents.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Surface Plasmon Resonance (SPR) Analysis of Binding Interactions of Inner-Ear Proteins.
Drescher DG, et al.
Methods in Molecular Biology, 1427, 165-165 (2016)
Co-purification and direct interaction of Ras with caveolin, an integral membrane protein of caveolae microdomains. Detergent-free purification of caveolae microdomains.
Song KS, et al.
The Journal of Biological Chemistry, 271, 9690-9690 (1996)
Cu 2 O/NiO x/graphene oxide modified glassy carbon electrode for the enhanced electrochemical oxidation of reduced glutathione and nonenzyme glucose sensor.
Yuan B et al.
Electrochimica Acta, 104, 78-78 (2013)
Nitric oxide as an independent regulatory factor in regenerating rat liver.
Tuncyurek P, et al.
Acta Chirurgica Belgica, 106, 581-581 (2006)
Hee Woong Yang et al.
FEBS letters, 593(15), 2060-2068 (2019-06-11)
Ribosomal protein S3 (rpS3) has endonuclease activity for DNA repair. In particular, rpS3 cleaves the phosphodiester bonds of damaged DNA. In this study, we show that the repair domain of rpS3 spans amino acids 144-189. We fused rpS3 with the

Articles

Amino Acid Codon Wheel for fast RNA translation. Find which amino acid is translated from your RNA sequence quickly and easily.

Amino Acid Codon Wheel for fast RNA translation. Find which amino acid is translated from your RNA sequence quickly and easily.

Amino Acid Codon Wheel for fast RNA translation. Find which amino acid is translated from your RNA sequence quickly and easily.

Amino Acid Codon Wheel for fast RNA translation. Find which amino acid is translated from your RNA sequence quickly and easily.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service