Skip to Content
Merck
All Photos(3)

Key Documents

159417

Sigma-Aldrich

Hydroxylamine hydrochloride

ReagentPlus®, 99%

Synonym(s):

Hydroxylammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2OH · HCl
CAS Number:
Molecular Weight:
69.49
Beilstein:
3539763
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.21
grade:
reagent
form:
crystalline

grade

reagent

Quality Level

vapor pressure

0.001 hPa ( 50 °C)

product line

ReagentPlus®

Assay

99%

form

crystalline

technique(s)

inhibition assay: suitable

pH

2.5-3.5 (20 °C, 50 g/L)

mp

155-157 °C (dec.) (lit.)

density

1.67 g/mL at 25 °C (lit.)

SMILES string

Cl.NO

InChI

1S/ClH.H3NO/c;1-2/h1H;2H,1H2

InChI key

WTDHULULXKLSOZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Hydroxylamine hydrochloride (Hydroxylammonium chloride) participates in the synthesis of 1,2,4-oxadiazoles, secondary amides and tertiary amides.
Hydroxylamine hydrochloride (NH2OH.HCl) is a hygroscopic salt widely used as a reactant in electrophilic substitution reactions, oxidation, and reduction reactions. It is also used in the synthesis of nitriles, pyrazoles, isoxazoles, nitrones, and pyridine.

Application

Hydroxylamine hydrochloride can be used as a reactant:
  • in the synthesis of primary amides from aldehydes in the presence of cesium carbonate (Cs2CO3) as a catalyst.
  • in the conversion of alicyclic /aliphatic carbonyl compounds and the aromatic aldehydes into corresponding oximes.
  • in the one-pot synthesis of nitriles from aldehydes in the presence of sodium sulfate (anhyd) and sodium bicarbonate catalysts.
  • It can also be used as a reducing agent in the preparation of single-layer reduced graphene oxide (RGO) sheets and films.

Biochem/physiol Actions

MAO inhibitor; inhibits platelet aggregation.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Carc. 2 - Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Oral

Target Organs

spleen

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Susie Suh et al.
Nature biomedical engineering, 5(2), 169-178 (2020-10-21)
Cytosine base editors and adenine base editors (ABEs) can correct point mutations predictably and independent of Cas9-induced double-stranded DNA breaks (which causes substantial indel formation) and homology-directed repair (which typically leads to low editing efficiency). Here, we show, in adult
Dongwoo Kim et al.
Analytical chemistry, 81(21), 9183-9187 (2009-10-31)
We report the use of electroless gold deposition as a light scattering signal enhancer in a multiplexed, microarray-based scanometric immunoassay using gold nanoparticle probes. The use of gold development results in greater signal enhancement than the typical silver development, and
Braedon McDonald et al.
Cell host & microbe, 28(5), 660-668 (2020-08-19)
Eradication of pathogens from the bloodstream is critical to prevent disseminated infections and sepsis. Kupffer cells in the liver form an intravascular firewall that captures and clears pathogens from the blood. Here, we show that the catching and killing of
Magnesia-supported hydroxylamine hydrochloride in the presence of sodium carbonate as an efficient reagent for the synthesis of 1, 2, 4-oxadiazoles from nitriles.
Kaboudin B and Saadati F.
Tetrahedron Letters, 48(16), 2829-2832 (2007)
Jian-Wei Hao et al.
Nature communications, 11(1), 4765-4765 (2020-09-23)
Fatty acids (FAs) are essential nutrients, but how they are transported into cells remains unclear. Here, we show that FAs trigger caveolae-dependent CD36 internalization, which in turn delivers FAs into adipocytes. During the process, binding of FAs to CD36 activates

Articles

Optimize serum samples for LC-MS/MS-based free testosterone determination using Supel™ BioSPME 96-pin device and validated equilibrium dialysis methods.

Optimize serum samples for LC-MS/MS-based free testosterone determination using Supel™ BioSPME 96-pin device and validated equilibrium dialysis methods.

Optimize serum samples for LC-MS/MS-based free testosterone determination using Supel™ BioSPME 96-pin device and validated equilibrium dialysis methods.

Optimize serum samples for LC-MS/MS-based free testosterone determination using Supel™ BioSPME 96-pin device and validated equilibrium dialysis methods.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service