Skip to Content
Merck
All Photos(1)

Key Documents

B2517

Sigma-Aldrich

6-Biopterin

≥97%

Synonym(s):

(1’R,2’S)-Biopterin, 2-Amino-4-hydroxy-6-(1,2-dihydroxypropyl)pteridine, L-erythro-6-(1,2-Dihydroxypropyl)pterin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11N5O3
CAS Number:
Molecular Weight:
237.22
Beilstein:
87860
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

Assay

≥97%

form

powder

solubility

1 M NaOH: soluble-10 mg/mL, clear, colorless to light greenish-yellow

storage temp.

2-8°C

SMILES string

C[C@H](O)[C@H](O)c1cnc2NC(N)=NC(=O)c2n1

InChI

1S/C9H11N5O3/c1-3(15)6(16)4-2-11-7-5(12-4)8(17)14-9(10)13-7/h2-3,6,15-16H,1H3,(H3,10,11,13,14,17)/t3-,6-/m0/s1

InChI key

LHQIJBMDNUYRAM-DZSWIPIPSA-N

Looking for similar products? Visit Product Comparison Guide

Application

6-Biopterin is an oxidation product from the melanogenesis regulating cofactor (6R)5,6,7,8 tetrahydrobiopterin (6-BH4). In vitro, 6-Biopterin is cytotoxic to melanocytes. However, in vivo, 6-Biopterin is rapidly and sequentially reduced back to 6-GH4 via q-BH2. Recent studies suggest that 6-Biopterin among other pteridines may be a useful non-invasive biomarker for cancer.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Siew Ling Choy et al.
Scientific reports, 7(1), 9472-9472 (2017-08-27)
Intracellular pathogens belonging to the genus Leishmania have developed effective strategies that enable them to survive within host immune cells. Immunostimulatory compounds that counteract such immunological escape mechanisms represent promising treatment options for diseases. Here, we demonstrate that a lipopeptidephosphoglycan
Sanjeewa Gamagedara et al.
Clinica chimica acta; international journal of clinical chemistry, 412(1-2), 120-128 (2010-09-28)
Biomarkers are good potential tools for early cancer diagnosis. Here we have analyzed eight different pteridines in the urine samples of cancer patients and compared them with samples from healthy subjects. Pteridines are important cofactors in the process of cell
Abulizi Abudukadier et al.
Diabetes, 62(9), 3033-3043 (2013-05-08)
Endothelial nitric oxide synthase (eNOS) dysfunction induces insulin resistance and glucose intolerance. Tetrahydrobiopterin (BH4) is an essential cofactor of eNOS that regulates eNOS activity. In the diabetic state, BH4 is oxidized to 7,8-dihydrobiopterin, which leads to eNOS dysfunction owing to
Jarl Underhaug et al.
Current topics in medicinal chemistry, 12(22), 2534-2545 (2013-01-24)
Phenylketonuria (PKU) is a loss-of-function inborn error of metabolism. As many other inherited diseases the main pathologic mechanism in PKU is an enhanced tendency of the mutant phenylalanine hydroxylase (PAH) to misfold and undergo ubiquitin-dependent degradation. Recent alternative approaches with
Suzanne V Saenko et al.
BMC biology, 11, 105-105 (2013-10-09)
Color traits in animals play crucial roles in thermoregulation, photoprotection, camouflage, and visual communication, and are amenable to objective quantification and modeling. However, the extensive variation in non-melanic pigments and structural colors in squamate reptiles has been largely disregarded. Here

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service