Skip to Content
Merck
All Photos(3)

Key Documents

H9039

Sigma-Aldrich

Hemin

from bovine, ≥90%

Synonym(s):

Chloro(protoporphyrinato)iron(III), Chlorohemin, Chloroprotoporphyrin IX iron(III), Ferriprotoporphyrin IX chloride, Hemin(chloride)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H32ClFeN4O4
CAS Number:
Molecular Weight:
651.94
Beilstein:
5229914
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.61

biological source

bovine

Quality Level

Assay

≥90%

form

powder

technique(s)

toxicology assay: suitable

solubility

1.4 M NH4OH: soluble 25 mg/mL

storage temp.

2-8°C

SMILES string

Cc1c(CCC(O)=O)c2cc3c(CCC(O)=O)c(C)c4cc5nc(cc6c(C)c(C=C)c(cc1n2)n6[Fe](Cl)n34)c(C=C)c5C

InChI

1S/C34H34N4O4.ClH.Fe/c1-7-21-17(3)25-13-26-19(5)23(9-11-33(39)40)31(37-26)16-32-24(10-12-34(41)42)20(6)28(38-32)15-30-22(8-2)18(4)27(36-30)14-29(21)35-25;;/h7-8,13-16H,1-2,9-12H2,3-6H3,(H4,35,36,37,38,39,40,41,42);1H;/q;;+3/p-3/b25-13-,26-13-,27-14-,28-15-,29-14-,30-15-,31-16-,32-16-;;

InChI key

BTIJJDXEELBZFS-HXFTUNQESA-K

Looking for similar products? Visit Product Comparison Guide

General description

Heme is a small molecule present either free or bound to hemoglobin in the bloodstream of mammals. This is an alternative source of iron within the host that contains a porphyrin ring containing a Fe2+ ion called hemin.

Application

Hemin was used to study conjugation, characterization and toxicity of lipophosphoglycan-polyacrylic acid conjugate for vaccination against leishmaniasis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Post-embryonic development in Caenorhabditis elegans.
D L Riddle
International journal for parasitology, 17(1), 223-231 (1987-02-01)
Xiaoyu Shi et al.
Molecular microbiology, 114(3), 454-467 (2020-05-21)
Reticulon and REEP family of proteins stabilize the high curvature of endoplasmic reticulum (ER) tubules. Plasmodium berghei Yop1 (PbYop1) is a REEP5 homolog in Plasmodium. Here, we characterize its function using a gene-knockout (Pbyop1∆). Pbyop1∆ asexual stage parasites display abnormal
Rodgoun Attarian et al.
mBio, 9(6) (2018-12-06)
The acquisition of iron and the maintenance of iron homeostasis are important aspects of virulence for the pathogenic fungus Cryptococcus neoformans In this study, we characterized the role of the monothiol glutaredoxin Grx4 in iron homeostasis and virulence in C.
Alberto J Ruiz et al.
Journal of biomedical optics, 25(5), 1-15 (2020-05-23)
Expanded use of fluorescence-guided surgery with devices approved for use with indocyanine green (ICG) has led to a range of commercial systems available. There is a compelling need to be able to independently characterize system performance and allow for cross-system
Murat Topuzogullari et al.
Journal of biomedical science, 20, 35-35 (2013-06-05)
Research on the conjugates of synthetic polyelectrolytes with antigenic molecules, such as proteins, peptides, or carbohydrates, is an attractive area due to their highly immunogenic character in comparison to classical adjuvants. For example, polyacrylic acid (PAA) is a weak polyelectrolyte

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service