Skip to Content
Merck
All Photos(4)

Key Documents

A7420

Sigma-Aldrich

Amyloglucosidase from Aspergillus niger

greener alternative

lyophilized powder, 30-60 units/mg protein (biuret), ≤0.02% glucose

Synonym(s):

1,4-α-D-Glucan glucohydrolase, Exo-1,4-α-glucosidase, Glucoamylase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

lyophilized powder

specific activity

30-60 units/mg protein (biuret)

composition

protein, ≥80%

greener alternative product characteristics

Waste Prevention
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

≤0.02% glucose

greener alternative category

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency and waste prevention when used in starch ethanol research. For more information see the article in biofiles.

Application

Amyloglucosidase from Aspergillus niger is used to hydrolyze α-D-glucosides. It may be used in the brewing of beer and in the production of bread and juices. Amyloglucosidase, from Sigma, has been used to hydrolyze glycogen into glucose monomers in order to study lipid accumulation in skeletal muscle .

Biochem/physiol Actions

Amyloglucosidase from Aspergillus niger is capable of hydrolyzing the α-D-(1-4), the α-D-(1-6), and the α-D-(1-3) glucosidic bonds of oligosaccharides. Amyloglucosidase is an extracellular enzyme that converts starch to dextrins and glucose. The enzyme is used in the starch-processing industry in the commercial production of D-glucose from corn syrup.

Unit Definition

One unit will liberate 1.0 mg of glucose from starch in 3 min at pH 4.5 at 55 °C.

Physical form

Lyophilized powder containing less than 0.02% glucose

inhibitor

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Youngmi Kim et al.
Bioresource technology, 99(12), 5165-5176 (2007-11-09)
DDGS and wet distillers' grains are the major co-products of the dry grind ethanol facilities. As they are mainly used as animal feed, a typical compositional analysis of the DDGS and wet distillers' grains mainly focuses on defining the feedstock's
Yi-Chen Lin et al.
PloS one, 12(5), e0177115-e0177115 (2017-05-05)
It has been proposed that malto-oligosaccharides (MOSs) are possibly recycled back into amylopectin biosynthesis via the sequential reactions catalyzed by plastidial α-glucan phosphorylase 1 (Pho1) and disproportionating enzyme 1 (Dpe1). In the present study, the reciprocal co-immunoprecipitation experiments using specific
Alisa K Manning et al.
Frontiers in genetics, 11, 615-615 (2020-08-06)
Causal transcripts at genomic loci associated with type 2 diabetes (T2D) are mostly unknown. The chr8p23.1 variant rs4841132, associated with an insulin-resistant diabetes risk phenotype, lies in the second exon of a long non-coding RNA (lncRNA) gene, LOC157273, located 175
Lizanne Janssens et al.
Aquatic toxicology (Amsterdam, Netherlands), 193, 210-216 (2017-11-04)
Pesticides are causing strong decreases in aquatic biodiversity at concentrations assumed safe by legislation. One reason for the failing risk assessment may be strong differences in the toxicity of the active ingredient of pesticides and their commercial formulations. Sublethal effects
Jaetaek Kim et al.
Molecular endocrinology (Baltimore, Md.), 22(11), 2531-2543 (2008-09-20)
The receptors for IGF-I (IGF-IR) and insulin (IR) have been implicated in physiological cardiac growth, but it is unknown whether IGF-IR or IR signaling are critically required. We generated mice with cardiomyocyte-specific knockout of IGF-IR (CIGF1RKO) and compared them with

Protocols

This procedure may be used for the determination of Amyloglucosidase activity using starch as the substrate.

This procedure may be used for the determination of Amyloglucosidase activity using starch as the substrate.

This procedure may be used for the determination of Amyloglucosidase activity using starch as the substrate.

This procedure may be used for the determination of Amyloglucosidase activity using starch as the substrate.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service