Skip to Content
Merck

Skip To

ROAMYGL

Roche

Amyloglucosidase

from Aspergillus niger

Synonym(s):

Glucoamylase, disaccharidase-type-α-D-glucosidase

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352204
EC Number:
NACRES:
NA.54
Specific activity:
~14 units/mg protein (At 25 °C with glycogen as the substrate; standardized with BSA.)
Biological source:
Aspergillus niger
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

Aspergillus niger

form

suspension

specific activity

~14 units/mg protein (At 25 °C with glycogen as the substrate; standardized with BSA.)

mol wt

Mr 97 kDa

packaging

pkg of 10 mL (10102857001 [100 mg])

manufacturer/tradename

Roche

parameter

55 °C optimum reaction temp.

optimum pH

4.6-4.8

storage temp.

2-8°C

General description

Amyloglucosidase is synthesized by several Aspergillus genus species. It is a disaccharidase–type α-glucosidase. This enzyme is an exo-enzyme and one of the major industrial enzymes. The stability of amyloglucosidase can be increased by immobilization.

Application

Amyloglucosidase from Aspergillus niger can be used for the hydrolyzation of terminal α1,4- and α1,6-glucosidic bonds (glucose-glucose bonds) in polysaccharides (e.g., starch, dextrins, glycogen), removing glucose units sequentially from the non-reducing end of the molecule. The enzyme will also cleave maltose and maltosides (maltotriose, maltotetraose, etc.).[1]

Biochem/physiol Actions

Amyloglucosidase from Aspergillus niger is capable of hydrolyzing the α-D-(1-4), the α-D-(1-6), and the α-D-(1-3) glucosidic bonds of oligosaccharides.[2] Amyloglucosidase is an extracellular enzyme that converts starch to dextrins and glucose. The enzyme is used in the starch-processing industry for the commercial production of D-glucose from corn syrups.[3][3]
Cleaves terminal glucoses that are α1,4- or α1,6-linked to an oligo- or polysaccharide of multiple glucose units. The product is D-glucose.
Heat inactivation: Heat inactivation is recommended at 80 °C for 45 minutes, followed by rapidly cooling down.

Physical form

Suspension in 3.2 M ammonium sulfate solution, pH approximately 6

Other Notes

For life science research only. Not for use in diagnostic procedures.
Unit Conversion: One unit (+25 °C; glycogen as substrate) corresponds to 8.6 U (+60 °C; starch as substrate).

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
ROAMYGLLA7095A1602
biological source

Aspergillus niger

biological source

Aspergillus niger

biological source

Aspergillus niger

biological source

-

specific activity

~14 units/mg protein (At 25 °C with glycogen as the substrate; standardized with BSA.)

specific activity

6 U/mg (approximately 6 U/mg lyophilizate at +25°C with glycogen as the substrate), ~6 units/mg protein (At 25 °C with glycogen as the substrate.)

specific activity

≥260 U/mL

specific activity

≥40 units/mg protein

form

suspension

form

lyophilized

form

aqueous solution

form

ammonium sulfate suspension

optimum pH

4.6-4.8

optimum pH

4.6-4.8

optimum pH

-

optimum pH

-

mol wt

Mr 97 kDa

mol wt

97 kDa

mol wt

-

mol wt

-

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C


Still not finding the right product?

Explore all of our products under Amyloglucosidase


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

does not flash

flash_point_c

does not flash



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library