Skip to Content
Merck
All Photos(2)

Key Documents

10113

Sigma-Aldrich

Amyloglucosidase from Aspergillus niger

greener alternative

powder, white, ~120 U/mg

Synonym(s):

1,4-α-D-Glucan glucohydrolase, Exo-1,4-α-glucosidase, Glucoamylase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

powder

specific activity

~120 U/mg

mol wt

Mr ~97000

greener alternative product characteristics

Waste Prevention
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

color

white

foreign activity

protease ≤0.1%

greener alternative category

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency and waste prevention when used in starch hydrolysis research. For more information see the article in biofiles.

Unit Definition

1 U corresponds to the amount of enzyme which liberates 1 μmol glucose per minute at pH 4.8 and 60 °C (starch, Cat. No. 85642, as substrate).

Other Notes

Synthesis of hetero-oligosaccharides by glucoamylase in reverse.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R.A. Rastall et al.
Biotechnology Letters, 13, 501-501 (1991)
Yuping Chen et al.
Bio-protocol, 7(13), e2371-e2371 (2017-07-05)
Organisms store carbohydrates in several forms. In yeast, carbohydrates are stored in glycogen (a multi-branched polysaccharide) and in trehalose (a disaccharide). As in other organisms, the amount of stored carbohydrate varies dramatically with physiological state, and accordingly, an assay of
Kulwa F Miraji et al.
Food & function, 12(16), 7527-7545 (2021-07-07)
Immature grain represents a precious nutritional source in many rural Africa areas. To optimize processing of immature rice into pepeta (a traditional rice-flakes produced from immature rice grains), immature rice (TXD306 variety) harvested at 18 and 26 days after 50%
Tiziana Ventriglia et al.
Plant physiology, 148(1), 65-76 (2008-07-11)
ADP-glucose (Glc) pyrophosphorylase (ADP-Glc PPase) catalyzes the first committed step in starch biosynthesis. Higher plant ADP-Glc PPase is a heterotetramer (alpha(2)beta(2)) consisting of two small and two large subunits. There is increasing evidence that suggests that catalytic and regulatory properties
Thomas Hundsberger et al.
Journal of neurology, 260(9), 2279-2285 (2013-06-12)
Glycogen storage disease type II is a rare multi-systemic disorder characterised by an intracellular accumulation of glycogen due a mutation in the acid alpha glucosidase (GAA) gene. The level of residual enzyme activity, the genotype and other yet unknown factors

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service