Skip to Content
Merck
All Photos(1)

Key Documents

57620

Sigma-Aldrich

Inulinase from Aspergillus niger

lyophilized, powder, brown-gray, ~25 U/mg

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

Aspergillus niger

form

powder

quality

lyophilized

specific activity

~25 U/mg

color

brown-gray

storage temp.

2-8°C

General description

Inulinases are classified as exoinulinases and endoinulinases.

Application

Inulinase from Aspergillus niger has been used as a glycoside hydrolase (GH) to study its potential to disperse bacteria (P. aeruginosa and S. aureus) from microbial biofilms using a polymicrobial well-plate model.

Biochem/physiol Actions

Inulinase hydrolyses inulin to produce oligosaccharides and liberate fructose. It also splits the terminal fructose units into sucrose and raffinose.
Inulinase is used in high-fructose syrup, which has therapeutic applications. Inulinase is reported to have high activity toward sucrose.

Unit Definition

1 U corresponds to the amount of enzyme which releases 1 μmol of reducing sugar (measured as fructose) per minute at pH 4.1 and 37°C from inulin

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R. Azhari et al.
Biotechnology and Applied Biochemistry, 11, 105-105 (1989)
Wen Wan et al.
Applied microbiology and biotechnology, 91(3), 789-798 (2011-06-10)
Easy and low-cost protein purification methods for the mass production of commonly used enzymes that play important roles in biotechnology are in high demand. In this study, we developed a fast, low-cost recombinant protein purification system in the methylotrophic yeast
Takahiro Rikita et al.
Analytical biochemistry, 426(1), 24-26 (2012-04-10)
Inulin is a polysaccharide composed mainly of D-fructose units and is the most reliable indicator of the glomerular filtration rate. We have proposed an inulin detection method that involves the hydrolysis of inulin to D-fructose using inulinase and the selective
Pierre Delanaye et al.
Clinica chimica acta; international journal of clinical chemistry, 413(5-6), 556-560 (2011-12-14)
Among issues susceptible to hamper a reliable measurement of inulin clearance, those regarding the dosage of inulin are largely neglected. We have compared the analytical performances of 2 commonly used methods of inulin dosage (one "acid" and one "enzymatic" method)
Gonçalo L M Santa et al.
Applied biochemistry and biotechnology, 165(1), 1-12 (2011-03-30)
The present work aims to provide the basic characterization of sol-gel immobilized inulinase, a biocatalyst configuration yet unexploited, using as model system the hydrolysis of inulin to fructose. Porous xerogel particles with dimensions in slight excess of 10 μm were obtained

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service