90A1
Leukocyte α-Naphthyl Acetate (Non-Specific Esterase) Kit
Select reagents packaged in gelatin capsules.
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
Quality Level
form
liquid
shelf life
Expiry date on the label.
IVD
for in vitro diagnostic use
application(s)
hematology
histology
shipped in
wet ice
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Esterases are ubiquitous in nature and encompass a series of different enzymes acting upon select substrates. According to the methodology, α-naphthyl acetate is enzymatically hydrolyzed, liberating a free naphthol compound. This then couples with a diazonium compound, forming black colored deposits at sites of non-specific esterase activity. This enzyme is detected primarily in monocytes, macrophages and histocytes, and is normally absent in granulocytes. Lymphocytes may occasionally exhibit enzyme activity. A sodium fluoride inhibition procedure may be run to demonstrate monocyte inhibition. The procedure is run at 37 °C and may be utilized with blood, bone marrow films, tissue-touch preparations, cytocentrifuge preparations and frozen sections. The enzyme does not survive paraffin processing.
Intended for the cytologic demonstration of a-naphthyl acetate esterase in blood, bone marrow films or tissue touch preparations.
Kit Components Only
Product No.
Description
- α-Naphthyl acetate (90-6) 10 x 20
- Citrate solution (386-1) 20 mL
- Ethylene glycol monomethyl ether 25 mL
- Fast Blue RR Salt (FBS-25) 10 caps
- Mayer's Hematoxylin Solution (MHS-1) 100 mL
- TRIZMAL™ 7.6 Concentrate 50 mL
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Met. Corr. 1 - Repr. 1B - Skin Corr. 1B - STOT RE 2 - STOT SE 1
Target Organs
Immune system, thymus
Storage Class Code
3 - Flammable liquids
Flash Point(F)
104.0 °F - closed cup
Flash Point(C)
40 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service