Skip to Content
Merck
All Photos(5)

Documents

D9805

Sigma-Aldrich

Fast Blue B Salt

Dye content ~95 %, Powder

Synonym(s):

o-Dianisidine bis(diazotized) zinc double salt, Azoic Diazo No. 48, DBB, Diazo Blue B, Naphthanil Diazo Blue B

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C14H12N4O2Cl2 · ZnCl2
CAS Number:
Molecular Weight:
475.47
Colour Index Number:
37235
Beilstein:
1499067
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

product name

Fast Blue B Salt, Dye content ~95 %

description

Technical grade

Quality Level

form

powder

composition

Dye content, ~95%

color

yellow to dark brown

mp

>300 °C (lit.)

solubility

H2O: 1 mg/mL

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-8°C

SMILES string

[Cl-].[Cl-].Cl[Zn]Cl.COc1cc(ccc1[N+]#N)-c2ccc([N+]#N)c(OC)c2

InChI

1S/C14H12N4O2.4ClH.Zn/c1-19-13-7-9(3-5-11(13)17-15)10-4-6-12(18-16)14(8-10)20-2;;;;;/h3-8H,1-2H3;4*1H;/q+2;;;;;+2/p-4

InChI key

GPPKNJIWDULNQH-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Application

  • Fast Blue B has been used in the acid phosphatase test in Clostridium perfringens.
  • It has been used as a spraying agent in thin layer chromatography for the identification of separated components.
  • It has also been used for the determination of the lipase activity.

Biochem/physiol Actions

Fast blue B is used for the determination of acetylcholinesterase activity. Acetylcholinesterase converts 1-naphthyl acetate to 1-naphthol, which reacts with fast blue B to form a purple-coloured diazonium dye. Similarly, fast blue B is also used for α- and β-glucosidase activity.

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Self-react. C

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C N Kennett et al.
Journal of clinical periodontology, 24(6), 424-431 (1997-06-01)
Gingival crevicular fluid (GCF) was collected from chronic periodontitis patients using plastic micropipettes and coverslip smears stained with antibodies for leukocyte markers and Toluidine Blue for mast cells. The smears consisted of 70-80% granulocytes, 10-20% monocytes/macrophages, 5% mast cells and
Overcoming the bell-shaped dose-response of cannabidiol by using Cannabis extract enriched in cannabidiol.
Gallily R, et al.
Pharmacology & Pharmacy, 6, 75-75 (2015)
Dong Ho Kang et al.
Polymers, 12(11) (2020-11-18)
In this study, composite anion exchange membranes (AEMs) were synthesized by cross-linking poly(phenylene oxide) (PPO) with cellulose functionalized by 1,4-diazabicyclo[2.2.2]-octane (DABCO) or di-guanidine (DG). The structural and morphological characteristics of the synthesized AEMs were characterized by FTIR, 1H-NMR, SEM, TEM
Partial characterization of pyloric-duodenal lipase of gilthead seabream (Sparus aurata).
Nolasco H, et al.
Fish Physiology and Biochemistry, 37, 43-43 (2011)
Evaluation of acid phosphatase as a confirmation test for Clostridium perfringens isolated from water.
Sartory DP, et al.
Letters in Applied Microbiology, 42, 418-418 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service