Skip to Content
Merck
All Photos(1)

Key Documents

68059

Sigma-Aldrich

FITC-CM-Dextran

average mol wt 4,000

Synonym(s):

FITC-CM-Dextran 4, Fluorescein isothiocyanate–Carboxymethyl–Dextran, Fluorescein isothiocyanate–Carboxymethyl–Dextran

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352116
NACRES:
NA.32

biological source

synthetic

conjugate

FITC conjugate

form

powder or crystals

mol wt

average mol wt 4,000

composition

carboxymethyl content, 3-7%

extent of labeling

0.001-0.020 mol/mol FITC glucose

technique(s)

permeability assay: suitable

fluorescence

λex 493 nm; λem 517 nm±5 nm in 0.1 M Tris pH 8.0

Looking for similar products? Visit Product Comparison Guide

General description

FITC-CM-Dextran, also known as Fluorescein isothiocyanate–Carboxymethyl–Dextran, is an anionic fluorescent probe. The carboxyl group provides the negative charge, which improves the overall permeability of the molecule. It is supplied as a yellow powder that is freely soluble in water and electrolyte solutions. FITC-CM-Dextran is insoluble in most organic samples such as ethanol, methanol, acetone, and chloroform. FITC-CM-Dextran is manufactured by reacting selected dextran fractions with an activated carboxymethyl derivative in alkali, whereby O-carboxymethyl groups are introduced along the dextran chain.

Application

FITC-CM-dextran is used to study drug intake and delivery mechanisms in HeLa cell lines. FITC-CM-Dextran was chosen due to its high water solubility and negative charge. It is also a suitable molecule to be used in encapsulation studies for polymersomal drug delivery systems or Ps DDS. FITC-CM-Dextran is a biocompatible molecule used as a starting material in several pharmaceutical and diagnostic applications.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stefanie J Anderson et al.
Comparative medicine, 69(1), 4-15 (2018-12-14)
Here we characterized the murine dextran sulfate sodium (DSS) model of acute colitis. Specifically, we evaluated azithromycin and metronidazole treatment regimens to assess their effects on animal wellbeing, pathologic changes, barrier function, cytokine and chemokine profiles, and neutrophil migration in
Yuxin Leng et al.
PloS one, 9(10), e109350-e109350 (2014-10-09)
Intra-abdominal hypertension (IAH) is known as a common, serious complication in critically ill patients. Bacterial translocation and permeability changes are considered the pathophysiological bases for IAH-induced enterogenic endotoxemia and subsequent multiorgan failure. Nevertheless, the effects of slightly elevated intra-abdominal pressures
Chloe Martin et al.
International journal of pharmaceutics, 511(1), 570-578 (2016-07-28)
The need to develop a greater understanding of drug delivery systems has arisen through the development of alternative biological based therapeutics. Drug delivery systems need to adapt and respond to this increasing demand for cellular transportation of highly charged species.
Yuxin Leng et al.
Scientific reports, 6, 22814-22814 (2016-03-17)
Intra-abdominal hypertension (IAH) is a common and serious complication in critically ill patients for which there is no well-defined treatment strategy. Here, we explored the effect of IAH on multiple intestinal barriers and discussed whether the alteration in microflora provides
Carrier and dose effects on the pharmacokinetics of T-0128, a camptothecin analogue-carboxymethyl dextran conjugate, in non-tumor- and tumor-bearing rats.
Harada M, Murata J, Sakamura Y, et al.
J. Controlled Release, 71, 71-86 (2001)

Articles

Explore FITC-labelled polysaccharides and their properties, including impact of molecular weight on molecule dimensions.

Explore FITC-labelled polysaccharides and their properties, including impact of molecular weight on molecule dimensions.

Explore FITC-labelled polysaccharides and their properties, including impact of molecular weight on molecule dimensions.

Explore FITC-labelled polysaccharides and their properties, including impact of molecular weight on molecule dimensions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service