Skip to Content
Merck
All Photos(1)

Documents

T1037

Sigma-Aldrich

Tetramethylrhodamine isothiocyanate–Dextran

average mol wt 4,400

Synonym(s):

TRITC-Dextran

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

form

solid

mol wt

average mol wt 4,400

extent of labeling

~1 TRITC per 100 glucose residues

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application


  • Preconcentration and Separation of Mixed-Species Samples Near a Nano-Junction in a Convergent Microchannel.: This article explores the separation of mixed-species samples using a nano-junction, employing TRITC-Dextran to track macromolecular movement within microchannels (Chiu et al., 2015).

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masashi Fujita et al.
PloS one, 7(1), e30224-e30224 (2012-01-19)
In the two-cell stage embryos of Caenorhabditis elegans, the contact surface of the two blastomeres forms a curve that bulges from the AB blastomere to the P₁ blastomere. This curve is a consequence of the high intracellular hydrostatic pressure of
Zhen-Yan Xia et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 67, 153158-153158 (2020-01-31)
Background Shengui Sansheng Pulvis (SSP) has about 300 years history used for stroke treatment, and evidences suggest it has beneficial effects on neuro-angiogenesis and cerebral energy metabolic amelioration post-stroke. However, its protective action and mechanisms on blood-brain barrier (BBB) is
Gustavo Vasen et al.
Proceedings of the National Academy of Sciences of the United States of America, 117(12), 6580-6589 (2020-03-11)
Polarity decisions are central to many processes, including mitosis and chemotropism. In Saccharomyces cerevisiae, budding and mating projection (MP) formation use an overlapping system of cortical landmarks that converges on the small G protein Cdc42. However, pheromone-gradient sensing must override
Sara C Sebag et al.
Experimental cell research, 362(2), 400-411 (2017-12-12)
Mitochondria are increasingly recognized as key mediators of acute cellular stress responses in asthma. However, the distinct roles of regulators of mitochondrial physiology on allergic asthma phenotypes are currently unknown. The mitochondrial Ca2+ uniporter (MCU) resides in the inner mitochondrial
Elena Naumovska et al.
International journal of molecular sciences, 21(14) (2020-07-18)
Intestinal organoids have emerged as the new paradigm for modelling the healthy and diseased intestine with patient-relevant properties. In this study, we show directed differentiation of induced pluripotent stem cells towards intestinal-like phenotype within a microfluidic device. iPSCs are cultured

Articles

Explore FITC-labelled polysaccharides and their properties, including impact of molecular weight on molecule dimensions.

Explore FITC-labelled polysaccharides and their properties, including impact of molecular weight on molecule dimensions.

Explore FITC-labelled polysaccharides and their properties, including impact of molecular weight on molecule dimensions.

Explore FITC-labelled polysaccharides and their properties, including impact of molecular weight on molecule dimensions.

Related Content

Step-by-step protocol for generating apical-out human gut organoids for microbiome, ADME/Tox, viral and gastrointestinal related disease research. See the complete organoid culture protocol.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service