Skip to Content
Merck

Skip To

FD40S

Fluorescein isothiocyanate–dextran

average mol wt 40,000

Synonym(s):

FITC–Dextran

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352201
NACRES:
NA.25
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic (organic)

Quality Segment

conjugate

FITC conjugate

form

powder

mol wt

average mol wt 40,000

extent of labeling

0.003-0.020 mol FITC per mol glucose

color

yellow to dark orange

solubility

H2O: soluble 25 mg/mL, clear to slightly hazy, yellow to deep orange

storage temp.

2-8°C

General description

Dextran is a polymer of anhydroglucose. It is composed of approximately 95% α-D-(166) linkages. The remaining (163) linkages account for the branching of dextran. Native dextran has been found to have a molecular weight (MW) in the range of 9 million to 500 million. FITC-dextran has an excitation maximum of 490 nm. It has a pH range of 5 to 9.

Application

Fluorescein isothiocyanate–dextran (FITC-dextran, fluorescein-dextran, FITC) was used in the following processes:
  • Fluorescein-dextran microscopy.
  • To evaluate the in vivo pulmonary vascular permeability in polymorphonuclear leukocyte α-defensins expressing transgenic mice.
  • To quantify endocytic activity by flow cytometry.
  • To label α-lactalbumin and ribonuclease.
FITC-dextran is suitable for extensive microcirculation and cell permeability research utilizing microfluorimetry. It is also suitable for the study of plant cell wall porosity and capillary permeability.
Dextran labeled with fluorescein isothiocyanate for possible use in perfusion studies in animals.

Preparation Note

Sigma typically tests the solubility of FITC dextrans in water at concentrations at or above 25 mg/ml. Solutions should be protected from light. In vivo, FITC-dextran is stable for more than 24 hours.

Other Notes

Native dextran has been found to have a molecular weight (MW) in the range of 9 million to 500 million. Lower MW dextrans will exhibit slightly less branching and have a more narrow range of MW distribution. Dextrans with MW greater than 10,000 behave as if they are highly branched. As the MW increases, dextran molecules attain greater symmetry. Dextrans with MW of 2,000 to 10,000, exhibit the properties of an expandable coil. At MW below 2,000, dextran is more rod-like. The MW of dextran is measured by one or more of the following methods: low angle laser light scattering, size exclusion chromatography, copper-complexation and anthrone reagent colorometric reducing-end sugar determination and viscosity.
Similar to FD-40, but produced by Sigma.
To gain a comprehensive understanding of our extensive range of Dextrans for your research, we encourage you to visit our Carbohydrates Category page.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
FD20SFD20FD150S
description

average mol wt 40,000

description

average mol wt 20,000

description

average mol wt 20,000

description

average mol wt 150,000

biological source

synthetic (organic)

biological source

sugar beets root

biological source

bacterial (Leuconostoc mesenteroides)

biological source

synthetic

solubility

H2O: soluble 25 mg/mL, clear to slightly hazy, yellow to deep orange

solubility

water: 25 mg/mL

solubility

water: 25 mg/mL

solubility

H2O: 25 mg/mL, clear to slightly hazy, yellow-orange

Quality Level

200

Quality Level

200

Quality Level

100

Quality Level

200

form

powder

form

powder

form

powder

form

powder

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

color

yellow to dark orange

color

yellow to dark yellow

color

light yellow to dark orange

color

faint yellow to orange


Still not finding the right product?


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–3 of 3 Questions  
  1. Can I dissolve this FITC-dextran in PBS to a concentration of 10 mg/mL as a stock solution, and then aliquot into small tubes kept in a box sheltered from light at -40 degrees Celcius?

    1 answer
    1. This product is routinely tested for solubility at 25 mg/mL in water. Solubility or solution stability in PBS has not been tested. Solubility in PBS is expected, as it is a common buffer used in various protocols. The cited protocol recommends storage on ice or at 4 °C wrapped in aluminum foil. No specific time frame for storage is provided, and no information regarding storage at -40 °C exists.

      Koester ST, Li N, Lachance DM, Dey N. Marker-based assays for studying gut transit in gnotobiotic and conventional mouse models. STAR Protoc. 2021 Nov 5;2(4):100938. doi: 10.1016/j.xpro.2021.100938. PMID: 34806048; PMCID: PMC8585659.

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  3. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters