Skip to Content
Merck
All Photos(1)

Key Documents

Y0001416

Betamethasone dipropionate for peak identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Betamethasone 17,21-dipropionate, 9α-Fluoro-16β-methyl-11β,17α,21-trihydroxy-1,4-pregnadiene-3,20-dione 17,21-dipropionate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H37FO7
CAS Number:
Molecular Weight:
504.59
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

betamethasone

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[H][C@@]12C[C@H](C)[C@](OC(=O)CCl)(C(=O)COC(=O)CC)[C@@]1(C)C[C@H](O)C3(F)[C@@]2([H])CCC4=CC(=O)C=C[C@]34C

InChI

1S/C27H34ClFO7/c1-5-22(33)35-14-21(32)27(36-23(34)13-28)15(2)10-19-18-7-6-16-11-17(30)8-9-24(16,3)26(18,29)20(31)12-25(19,27)4/h8-9,11,15,18-20,31H,5-7,10,12-14H2,1-4H3/t15-,18-,19-,20-,24-,25-,26?,27-/m0/s1

InChI key

UOCNBCVPZWLUKX-HOBFILNXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Betamethasone dipropionate for peak identification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Betamethasone 17,21-dipropionate is a glucocorticoid with anti-inflammatory and immunosuppressive activity.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B - STOT RE 2

Target Organs

Liver,Kidney,Endocrine system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrew G Affleck et al.
Current medical research and opinion, 27(1), 269-284 (2010-12-15)
To compare the cost effectiveness of the two-compound formulation (TCF) calcipotriol plus betamethasone dipropionate gel used first-, second- or third-line to standard topical treatments for moderately severe scalp psoriasis from a Scottish NHS perspective. Treatment pathways for scalp psoriasis patients
Paul L McCormack
Drugs, 71(6), 709-730 (2011-04-21)
Calcipotriol/betamethasone dipropionate (calcipotriol 50 μg/g and betamethasone 0.5 mg/g) is a fixed-dose combination of a vitamin D(3) analogue and a corticosteroid indicated for the once-daily, topical treatment of psoriasis vulgaris of the trunk, limbs and scalp in adults. Both the ointment (Daivobet®;
Peter van de Kerkhof et al.
Current medical research and opinion, 27(1), 225-238 (2010-12-15)
The efficacy of the two-compound formulation (TCF) product containing calcipotriol and betamethasone dipropionate applied once daily in psoriasis has been demonstrated in phase III trials but no randomised clinical trial comparing all commonly used topical treatments exists. The aim of
M A Radtke et al.
Der Hautarzt; Zeitschrift fur Dermatologie, Venerologie, und verwandte Gebiete, 61(9), 770-775 (2010-07-21)
Scalp psoriasis has a significant psychosocial impact on individuals affecting their quality of life. It occurs in 50-80% of patients with psoriasis and may be the only affected area of the body. Current topical therapies for scalp psoriasis are difficult
F Henry et al.
Revue medicale de Liege, 60(11), 893-895 (2006-01-13)
The topical treatment of psoriasis benefits from the alternate use of dermocorticosteroids and vitamin D3 analogues. A new galenic formulation allows to combine them in a single application. Dovobet (LEO Pharma) ointment is the association of calcipotriol 50 microg/g with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service