Skip to Content
Merck

Skip To

1066009

USP

Betamethasone

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

9α-Fluoro-11β,17α,21-trihydroxy-16β-methylpregna-1,4-diene-3,20-dione, 9α-Fluoro-16β-methyl-11β,17α,21-trihydroxy-1,4-pregnadiene-3,20-dione, 9α-Fluoro-16β-methylprednisolone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C22H29FO5
CAS Number:
Molecular Weight:
392.46
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic

grade

pharmaceutical primary standard

agency

USP

vapor pressure

<0.0000001 kPa ( 25 °C)

API family

betamethasone

packaging

pkg of 200 mg

manufacturer/tradename

USP

color

white to off-white

mp

235-237 °C (lit.), 447.8-473 °F (231—245°C; decomposes)

solubility

acetone: sparingly soluble, chloroform: very slightly soluble, ethanol: sparingly soluble, ether: very slightly soluble, methanol: sparingly soluble, water: insoluble

density

0.305 g/cm3 at 25 °C (77°F)

application(s)

pharmaceutical
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[H][C@@]12C[C@H](C)[C@](O)(C(=O)CO)[C@@]1(C)C[C@H](O)[C@@]3(F)[C@@]2([H])CCC4=CC(=O)C=C[C@]34C

InChI

1S/C22H29FO5/c1-12-8-16-15-5-4-13-9-14(25)6-7-19(13,2)21(15,23)17(26)10-20(16,3)22(12,28)18(27)11-24/h6-7,9,12,15-17,24,26,28H,4-5,8,10-11H2,1-3H3/t12-,15-,16-,17-,19-,20-,21-,22-/m0/s1

InChI key

UREBDLICKHMUKA-DVTGEIKXSA-N

Gene Information

human ... NR3C1(2908)

General description

Find answers to frequently asked questions on USP products in USP FAQs.
Betamethasone, a synthetic glucocorticoid, belongs to the corticosteroid family of active pharmaceutical ingredients (APIs).
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Betamethasone USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Betamethasone Acetate
  • Betamethasone Oral Solution
  • Betamethasone Valerate Cream
  • Betamethasone Valerate Lotion
  • Betamethasone Valerate Ointment
  • Dexamethasone

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
BP575B700534166
description

United States Pharmacopeia (USP) Reference Standard

description

British Pharmacopoeia (BP) Reference Standard

description

≥98%

description

VETRANAL®, analytical standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

-

grade

analytical standard

manufacturer/tradename

USP

manufacturer/tradename

BP

manufacturer/tradename

-

manufacturer/tradename

-

format

neat

format

neat

format

-

format

neat

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical
pharmaceutical small molecule

application(s)

-

application(s)

clinical testing

API family

betamethasone

API family

betamethasone

API family

-

API family

-

vapor pressure

<0.0000001 kPa ( 25 °C)

vapor pressure

-

vapor pressure

-

vapor pressure

-


Still not finding the right product?

Explore all of our products under Betamethasone


Pictograms

Health hazard

Signal Word

Danger

Hazard Codes

Hazard Classifications

Repr. 1B - STOT RE 2

Target Organs

Liver,Kidney,Endocrine system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters